Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805427_g1
          See other SSRP1 GE Assays ›
          Gene Symbol:
          Ssrp1
          Gene Name:
          structure specific recognition protein 1
          Gene Aliases:
          C81323, Hmg1-rs1, Hmgi-rs3, Hmgox, T160
          Chromosome Location:
          Chr.2: 85037120 - 85047114 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805427_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ssrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136081.2 NP_001129553.1 8 - 9 1210 65
          NM_182990.4 NP_892035.2 8 - 9 1339 65
          GenBank mRNA AK146446.1 - 8 - 9 1262 65
          AK146585.1 - 8 - 9 1259 65
          AK146607.1 - 8 - 9 1242 65
          AK146691.1 - 8 - 9 1242 65
          AK147005.1 - 8 - 9 1270 65
          AK158552.1 - 8 - 9 1210 65
          BC004848.1 - 8 - 9 1206 65
          BC024835.1 - 5 - 6 709 65
          BC042502.1 - 8 - 9 1187 5005209 65
          S50213.1 - 7 - 8 1084 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001229.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ssrp1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA repair
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          biological_process
          DNA binding
          chromatin binding
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200009859.mm.1 TC0200001229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009859.mm.1 NM_001136081.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009859.mm.1 NM_182990.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009857.mm.1 TC0200001229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009857.mm.1 NM_001136081.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009857.mm.1 NM_182990.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0