Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00806354_m1
          See other RNMTL1 GE Assays ›
          Gene Symbol:
          Rnmtl1
          Gene Name:
          RNA methyltransferase like 1
          Gene Aliases:
          4833420N02Rik, AI255153, HC90
          Chromosome Location:
          Chr.11: 76243736 - 76250625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00806354_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnmtl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183263.5 NP_899086.2 2 - 3 592 108
          GenBank mRNA AK012200.1 - 2 - 3 566 108
          AK014739.1 - 2 - 3 558 108
          AK147805.1 - 2 - 3 573 108
          BC038288.1 - 2 - 3 541 108
          BC090975.1 - 2 - 3 564 6595681 108
          BC115622.1 - 2 - 3 318 40102799 108
          BC115623.1 - 2 - 3 318 40102801 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001097.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnmtl1

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          RNA processing
          biological_process
          methylation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          RNA binding
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930798 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 BC090975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK014739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK147805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 NM_183263.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK012200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 BC090975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK014739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK147805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 NM_183263.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401527 AK012200.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93982 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162521 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 BC090975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK014739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK147805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 NM_183263.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK012200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 BC090975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK014739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK147805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 NM_183263.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401527 AK012200.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162521 - -
          Whole Mouse Genome G4122A A_51_P401527 BC090975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK014739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK147805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 NM_183263.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK012200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 BC090975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK014739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK147805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 NM_183263.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401527 AK012200.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113192 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162521 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09260073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09260076 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09260086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09535282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09535272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09535270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378772 - -
          Affymetrix HT MG-430 PM Array Plate 1428766_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378768 - -
          Mouse Genome 430 2.0 Array 1428766_PM_at - -
          Mouse Genome 430 2.0 Array 1428766_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67390_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67390_at - -
          Affymetrix Mouse Clariom S Array PSR1100009983.mm.1 TC1100001097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009983.mm.1 NM_183263.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009983.mm.1 NM_183263.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009985.mm.1 TC1100001097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009985.mm.1 NM_183263.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009985.mm.1 NM_183263.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009980.mm.1 TC1100001097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009980.mm.1 NM_183263.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009980.mm.1 NM_183263.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009981.mm.1 TC1100001097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009981.mm.1 NM_183263.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009981.mm.1 NM_183263.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009982.mm.1 TC1100001097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009982.mm.1 NM_183263.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009982.mm.1 NM_183263.5 Refseq
          Mouse Expression Set 430 1428766_PM_at - -
          Mouse Expression Set 430 1428766_at - -
          Murine Genome U74v2 Set 161756_PM_at - -
          Murine Genome U74v2 Set 161756_at - -
          Murine Genome U74v2 Set 160722_PM_at - -
          Murine Genome U74v2 Set 160722_at - -
          Mouse Genome 430A 2.0 Array 1428766_at - -
          Mouse Genome 430A 2.0 Array 1428766_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428766_at - -
          Affymetrix HT Mouse Genome 430A Array 1428766_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 629558 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990072 - -
          Illumina mouse-6 v1.0 expression beadchip 1990072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715234 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715234 - -
          mouse-6 v1.1 (Illumina) scl41246.4.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41246.4.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2715234 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715234 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-fb85479f-282mh:80/100.66.130.62:80.
          git-commit: 1c69677c60948f80141da00d3c5091e873d99202
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline