Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809064_s1
          See other B130024G19RIK GE Assays ›
          Gene Symbol:
          B130024G19Rik Nr2f2
          Gene Name:
          RIKEN cDNA B130024G19 gene
          nuclear receptor subfamily 2, group F, member 2
          Gene Aliases:
          -
          2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2
          Chromosome Location:
          Chr.7: 70365383 - 70411146 on Build GRCm38
          Chromosome Location:
          Chr.7: 70351946 - 70366746 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00809064_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B130024G19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK051651.1 - 1 551 152
          Gene Symbol: Nr2f2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK012322.1 - 1 152
          AK012573.1 - 1 152

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177463 BC070425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177463 AK045070.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103380 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177463 BC070425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177463 AK045070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91823 BC070425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91823 AK045070.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06220792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06437215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06220801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06633426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06379239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05939006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06662952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06314268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06035300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06459943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06576948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06314269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06541393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25210998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25211049 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554131 - -
          Affymetrix HT MG-430 PM Array Plate 1436658_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10554129 - -
          Mouse Genome 430 2.0 Array 1431237_PM_at - -
          Mouse Genome 430 2.0 Array 1436658_at - -
          Mouse Genome 430 2.0 Array 1431237_at - -
          Mouse Genome 430 2.0 Array 1436658_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434198_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434198_PM_at - -
          Mouse Expression Set 430 1436658_PM_at - -
          Mouse Expression Set 430 1436658_at - -
          Mouse Expression Set 430 1431237_at - -
          Mouse Expression Set 430 1431237_PM_at - -
          Mouse Genome 430A 2.0 Array 1436658_at - -
          Mouse Genome 430A 2.0 Array 1436658_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436658_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436658_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 104570707 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232469 - -
          Illumina Sentrix mouse-6.v1 bead array 104570707 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104570707 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0