Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809562_s1
          See other TUBB2A GE Assays ›
          Gene Symbol:
          Tubb2a
          Gene Name:
          tubulin, beta 2A class IIA
          Gene Aliases:
          M(beta)2, Tubb2
          Chromosome Location:
          Chr.13: 34074278 - 34078008 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00809562_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubb2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009450.2 NP_033476.1 4 1474 74
          GenBank mRNA AK134545.1 - 4 1473 74
          BC003475.1 - 1 954 74
          BC055441.1 - 4 1464 4016867 74
          CB522913.1 - 1 288 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001779.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tubb2a

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          protein polymerization
          nucleotide binding
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936501 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936503 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490023 M28739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P490023 M28739.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347628 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490023 M28739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P490023 M28739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P621603 AK168908.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P621603 BC138935.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P621603 BC138936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P621603 NM_023716.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P621603 AK010786.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103659 AK134545.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103659 BC055441.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103659 CJ148662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103659 NM_009450.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490023 M28739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490023 M28739.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28936493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154967 - -
          Whole Mouse Genome G4122A A_51_P103659 AK134545.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103659 BC055441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103659 CJ148662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103659 NM_009450.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490023 M28739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490023 M28739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P621603 AK168908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P621603 BC138935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P621603 BC138936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P621603 NM_023716.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P621603 AK010786.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133727 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528958 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10529007 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10528966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936499 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936500 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28936501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28936533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11112385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10986834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10528965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10529000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10528962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10528951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10986840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11049834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10528959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399420 - -
          Affymetrix HT MG-430 PM Array Plate 1427347_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1427838_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10399419 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408610 - -
          Mouse Genome 430 2.0 Array 1427838_PM_at - -
          Mouse Genome 430 2.0 Array 1427347_s_at - -
          Mouse Genome 430 2.0 Array 1427838_at - -
          Mouse Genome 430 2.0 Array 1427347_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22151_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22151_at - -
          Affymetrix Mouse Clariom S Array PSR1300011109.mm.1 TC1300001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011109.mm.1 NM_009450.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011109.mm.1 NM_009450.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011108.mm.1 TC1300001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011108.mm.1 NM_009450.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011108.mm.1 NM_009450.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011111.mm.1 TC1300001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011111.mm.1 NM_009450.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011111.mm.1 NM_009450.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011110.mm.1 TC1300001779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011110.mm.1 NM_009450.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011110.mm.1 NM_009450.2 Refseq
          Mouse Expression Set 430 1427838_at - -
          Mouse Expression Set 430 1427347_s_at - -
          Mouse Expression Set 430 1427838_PM_at - -
          Mouse Expression Set 430 1427347_PM_s_at - -
          Murine Genome U74v2 Set 161614_r_at - -
          Murine Genome U74v2 Set 94835_f_at - -
          Murine Genome U74v2 Set 161614_PM_r_at - -
          Murine Genome U74v2 Set 94835_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1427838_at - -
          Mouse Genome 430A 2.0 Array 1427347_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427838_PM_at - -
          Mouse Genome 430A 2.0 Array 1427347_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427838_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427347_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427838_at - -
          Affymetrix HT Mouse Genome 430A Array 1427347_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 398481 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6940435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2485044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2485044 - -
          mouse-6 v1.1 (Illumina) scl44112.4_403-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44112.4_403-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2485044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2485044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline