Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00824486_g1
          See other PCDHB6 GE Assays ›
          Gene Symbol:
          Pcdhb6
          Gene Name:
          protocadherin beta 6
          Gene Aliases:
          Pcdhb5B, PcdhbF
          Chromosome Location:
          Chr.18: 37334028 - 37336346 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00824486_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053131.1 NP_444361.1 1 1221 127
          GenBank mRNA AK032193.1 - 834 127
          AY013788.1 - 1 1221 127
          BC150716.1 - 1 1303 9087627 127

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb6

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 BC150716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 AK032193.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 AY013788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 BC150716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 AK032193.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 AY013788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137996 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 BC150716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 AK032193.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 AY013788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 NM_053131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 BC150716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 AK032193.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 AY013788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 BC150716.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 AK032193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 AY013788.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 BC150716.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 AK032193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 AY013788.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31627903 - -
          Whole Mouse Genome G4122A A_51_P403413 BC150716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 AK032193.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 AY013788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 NM_053131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 BC150716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 AK032193.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 AY013788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403413 NM_053131.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101341 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13694500 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13694529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13694533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13694531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31627974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13694524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13988314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13694536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13694508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13694531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31627865 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455070 - -
          Affymetrix HT MG-430 PM Array Plate 1423014_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455069 - -
          Mouse Genome 430 2.0 Array 1423014_PM_at - -
          Mouse Genome 430 2.0 Array 1423014_at - -
          Mouse Expression Set 430 1423014_at - -
          Mouse Expression Set 430 1423014_PM_at - -
          Mouse Genome 430A 2.0 Array 1423014_PM_at - -
          Mouse Genome 430A 2.0 Array 1423014_at - -
          Affymetrix HT Mouse Genome 430A Array 1423014_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423014_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6420181 - -
          Illumina mouse-6 v1.0 expression beadchip 100060164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2497410 - -
          mouse-6 v1.1 (Illumina) scl0093877.2_98-S - -
          mouse-6 v1.1 (Illumina) scl52069.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420181 - -
          Illumina Sentrix mouse-6.v1 bead array 100060164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0093877.2_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100060164 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2497410 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0