Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833353_mH
          See other GSTA2 GE Assays ›
          Gene Symbol:
          Gsta2
          Gene Name:
          glutathione S-transferase, alpha 2 (Yc2)
          Gene Aliases:
          Gst2-2, Gstc-2, Gstc2
          Chromosome Location:
          Chr.9: 78331018 - 78347192 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833353_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsta2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008182.3 NP_032208.2 5 - 6 485 212
          GenBank mRNA AK002712.1 - 5 - 6 492 212
          BC061133.1 - 4 - 5 428 30303519 212
          J03958.1 - 5 - 6 500 212

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002719.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gsta2

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          glutathione transferase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170454 AK002712.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170454 BC061133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170454 NM_008182.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144370 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129934 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170454 AK002712.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170454 BC061133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170454 NM_008182.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659144 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01978986 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06853376 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174942 - -
          Whole Mouse Genome G4122A A_52_P458682 AK002712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458682 BC061133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458682 J03958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458682 NM_008182.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08160264 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08160261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07812718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26658994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26658960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26658959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26659107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07973364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07657488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07820492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07657489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08160264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07518732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07576722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07860703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07458605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07780479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595155 - -
          Affymetrix HT MG-430 PM Array Plate 1421040_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587331 - -
          Affymetrix Mouse Gene 1.1 ST Array 10587339 - -
          Affymetrix Mouse Gene 1.1 ST Array 10595148 - -
          Affymetrix Mouse Gene 1.1 ST Array 10587323 - -
          Mouse Genome 430 2.0 Array 1421041_s_at - -
          Mouse Genome 430 2.0 Array 1421041_PM_s_at - -
          Mouse Genome 430 2.0 Array 1421040_a_at - -
          Mouse Genome 430 2.0 Array 1421040_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14858_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14858_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900022438.mm.1 TC0900002719.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022438.mm.1 NM_008182.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022438.mm.1 NM_008182.3 Refseq
          Mouse Expression Set 430 1421041_PM_s_at - -
          Mouse Expression Set 430 1421040_a_at - -
          Mouse Expression Set 430 1421040_PM_a_at - -
          Mouse Expression Set 430 1421041_s_at - -
          Murine Genome U74v2 Set 101872_at - -
          Murine Genome U74v2 Set 101872_PM_at - -
          Mouse Genome 430A 2.0 Array 1421041_s_at - -
          Mouse Genome 430A 2.0 Array 1421040_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1421040_a_at - -
          Mouse Genome 430A 2.0 Array 1421041_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421040_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421040_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6180010 - -
          MouseWG-6 v2 Expression BeadChip 106770161 - -
          Illumina mouse-6 v1.0 expression beadchip 106770161 - -
          Illumina mouse-6 v1.0 expression beadchip 6180010 - -
          Illumina mouse-6 v1.0 expression beadchip 6550139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248849 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248781 - -
          mouse-6 v1.1 (Illumina) GI_6680118-S - -
          mouse-6 v1.1 (Illumina) scl00235501.1_123-S - -
          mouse-6 v1.1 (Illumina) scl0014858.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180010 - -
          Illumina Sentrix mouse-6.v1 bead array 106770161 - -
          Illumina Sentrix mouse-6.v1 bead array 6550139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00235501.1_123-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014858.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106770161 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248849 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248849 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix