Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834201_g1
          See other RPL9 GE Assays ›
          Gene Symbol:
          Rpl9
          Gene Name:
          ribosomal protein L9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 65388364 - 65391431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834201_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011292.2 NP_035422.1 7 - 8 648 63
          GenBank mRNA AF260271.1 - 6 - 7 618 63
          AK012331.1 - 7 - 8 618 63
          AK012445.1 - 7 - 8 618 63
          AK017388.1 - 7 - 8 618 63
          AK017427.1 - 7 - 8 618 63
          AK084278.1 - 7 - 8 619 63
          AK088166.1 - 7 - 8 618 63
          AK153562.1 - 7 - 8 618 63
          AK159980.1 - 7 - 8 618 63
          AK168326.1 - 7 - 8 616 63
          AK169217.1 - 7 - 8 616 63
          AK204218.1 - 3 - 4 221 63
          BC013165.1 - 6 - 7 608 2655358 63
          BC081435.1 - 6 - 7 594 6824107 63
          BC083166.1 - 6 - 7 599 6397429 63
          BC083329.1 - 6 - 7 597 6824346 63
          BC086937.1 - 7 - 8 615 6787671 63
          BC089319.1 - 6 - 7 597 6823996 63
          BC093526.1 - 6 - 7 597 5150493 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002484.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl9

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756087 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756084 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998825 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31210 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133891 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998825 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301734 BC089319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301734 BC093526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301734 U17331.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301734 U17332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301734 AK153562.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23756060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133891 - -
          Whole Mouse Genome G4122A A_51_P301734 BC089319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301734 BC093526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301734 U17331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301734 U17332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301734 AK153562.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128585 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109736 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04203195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04203215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04203213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04203226 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04203224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756087 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23756084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23756067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04488139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04488133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23756081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530197 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530196 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530195 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410507 - -
          Affymetrix HT MG-430 PM Array Plate 1436798_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443843_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10439402 - -
          Affymetrix Mouse Gene 1.1 ST Array 10472686 - -
          Affymetrix Mouse Gene 1.1 ST Array 10467088 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389229 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10530194 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410506 - -
          Affymetrix Mouse Gene 1.1 ST Array 10489049 - -
          Mouse Genome 430 2.0 Array 1436798_at - -
          Mouse Genome 430 2.0 Array 1443843_x_at - -
          Mouse Genome 430 2.0 Array 1416420_a_at - -
          Mouse Genome 430 2.0 Array 1436798_PM_at - -
          Mouse Genome 430 2.0 Array 1416420_PM_a_at - -
          Mouse Genome 430 2.0 Array 1443843_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0500022155.mm.1 TC0500002484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022155.mm.1 NM_011292.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022155.mm.1 NM_011292.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022156.mm.1 TC0500002484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022156.mm.1 NM_011292.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022156.mm.1 NM_011292.2 Refseq
          Mouse Expression Set 430 1416420_a_at - -
          Mouse Expression Set 430 1436798_PM_at - -
          Mouse Expression Set 430 1436798_at - -
          Mouse Expression Set 430 1416420_PM_a_at - -
          Mouse Expression Set 430 1443843_x_at - -
          Mouse Expression Set 430 1443843_PM_x_at - -
          Murine Genome U74v2 Set 167724_r_at - -
          Murine Genome U74v2 Set 167724_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1436798_at - -
          Mouse Genome 430A 2.0 Array 1416420_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1436798_PM_at - -
          Mouse Genome 430A 2.0 Array 1416420_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1436798_at - -
          Affymetrix HT Mouse Genome 430A Array 1436798_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443843_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443843_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102030270 - -
          Illumina mouse-6 v1.0 expression beadchip 102030270 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235171 - -
          mouse-6 v1.1 (Illumina) GI_14149646-S - -
          Illumina Sentrix mouse-6.v1 bead array 102030270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102030270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2851231 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2851231 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0