Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834357_g1
          See other EIF4A2 GE Assays ›
          Gene Symbol:
          Eif4a2
          Gene Name:
          eukaryotic translation initiation factor 4A2
          Gene Aliases:
          4833432N07Rik, BM-010, Ddx2b, Eif4, eIF-4A-II, eIF4A-II
          Chromosome Location:
          Chr.16: 23107468 - 23114132 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834357_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif4a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001123038.2 NP_001116510.1 3 - 4 529 99
          NM_013506.3 NP_038534.2 3 - 4 529 99
          NR_110335.1 3 - 4 529 99
          GenBank mRNA AK076509.1 - 3 - 4 240 99
          AK088669.1 - 3 - 4 241 99
          AK132969.1 - 3 - 4 240 99
          BC094422.1 - 3 - 4 225 6808404 99
          CF723811.1 - 3 - 4 421 99
          X12507.1 - 3 - 4 241 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000329.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif4a2

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational initiation
          regulation of gene expression
          RNA secondary structure unwinding
          negative regulation of RNA-directed 5'-3' RNA polymerase activity
          nucleotide binding
          nucleic acid binding
          RNA binding
          RNA helicase activity
          translation initiation factor activity
          helicase activity
          ATP binding
          hydrolase activity
          ATP hydrolysis activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600003288.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003288.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003288.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003288.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003272.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003272.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003272.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003272.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003272.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003277.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003277.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003277.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003277.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003277.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003279.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003279.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003279.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003279.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003279.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003287.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003287.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003287.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003287.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003293.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003293.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003293.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003293.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003293.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003285.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003285.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003285.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003285.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003262.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003262.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003262.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003262.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003262.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003286.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003286.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003286.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003286.mm.1 NR_110335.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003292.mm.1 TC1600000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003292.mm.1 NM_001123037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003292.mm.1 NM_001123038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003292.mm.1 NM_013506.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003292.mm.1 NR_110335.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 569766 - -
          ABI Mouse Genome Survey Microarray 895852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline