Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834592_g1
          See other MPC1 GE Assays ›
          Gene Symbol:
          Mpc1
          Gene Name:
          mitochondrial pyruvate carrier 1
          Gene Aliases:
          0610006G08Rik, 3830411I18Rik, Brp44l
          Chromosome Location:
          Chr.17: 8283813 - 8297667 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834592_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018819.4 NP_061289.1 3 - 4 250 85
          GenBank mRNA AF181116.1 - 3 - 4 206 85
          AK002241.1 - 3 - 4 251 85
          AK002266.1 - 3 - 4 229 85
          AK004013.1 - 3 - 4 206 85
          AK005348.1 - 3 - 4 242 85
          AK013391.1 - 3 - 4 251 85
          AK014435.1 - 3 - 4 480 85
          AK135943.1 - 3 - 4 221 85
          AK170514.1 - 3 - 4 267 85
          BC024365.1 - 3 - 4 212 4988147 85
          BC103767.1 - 3 - 4 253 30840199 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000085.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mpc1

          Gene Ontology Categories:

          Function(s) Process(es)

          pyruvate metabolic process
          transport
          mitochondrial pyruvate transmembrane transport
          pyruvate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957267 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957261 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957259 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957256 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962092 AK170514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962092 BC024365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962092 BC103767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962092 AK013391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962092 AK014421.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008422 AK043779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008422 AK170514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008422 BC103767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008422 AK013391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008422 AK014421.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48770 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121054 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962092 AK170514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962092 BC024365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962092 BC103767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962092 AK013391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962092 AK014421.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008422 AK043779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008422 AK170514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008422 BC103767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008422 AK013391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008422 AK014421.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444301 AK170514.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444301 BC024365.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444301 BC103767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444301 AK013391.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444301 AK014421.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09553433 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821646 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09553415 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821648 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09553421 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03821656 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121054 - -
          Whole Mouse Genome G4122A A_51_P444301 AK170514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444301 BC024365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444301 BC103767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444301 AK013391.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P444301 AK014421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595687 AK014435.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128619 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126427 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121054 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159723 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957261 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957258 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957257 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957267 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957259 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32957264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32957248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402649 - -
          Affymetrix HT MG-430 PM Array Plate 1453651_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10441642 - -
          Affymetrix Mouse Gene 1.1 ST Array 10402648 - -
          Mouse Genome 430 2.0 Array 1453651_a_at - -
          Mouse Genome 430 2.0 Array 1453651_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1700000681.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000681.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000681.mm.1 NM_018819.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000683.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000683.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000683.mm.1 NM_018819.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000685.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000685.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000685.mm.1 NM_018819.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000680.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000680.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000680.mm.1 NM_018819.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000684.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000684.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000684.mm.1 NM_018819.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000687.mm.1 TC1700000085.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000687.mm.1 NM_018819.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000687.mm.1 NM_018819.4 Refseq
          Mouse Expression Set 430 1453651_a_at - -
          Mouse Expression Set 430 1453651_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453651_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453651_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453651_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453651_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740838 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740838 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0