Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835386_gH
          See other RPS27L GE Assays ›
          Gene Symbol:
          Rps27l
          Gene Name:
          ribosomal protein S27-like
          Gene Aliases:
          1810034D23Rik
          Chromosome Location:
          Chr.9: 66946076 - 66949520 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835386_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps27l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026467.3 NP_080743.1 3 - 4 280 155
          GenBank mRNA AK007689.1 - 3 - 4 281 155
          AK011262.1 - 3 - 4 299 155
          AK151083.1 - 3 - 4 299 155
          BC058115.1 - 3 - 4 281 6407738 155

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000876.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps27l

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          translation
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          mitotic G1 DNA damage checkpoint signaling
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of translation
          RNA binding
          structural constituent of ribosome
          translation activator activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596134 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596133 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596130 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596132 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK011262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK151083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 BC058115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 NM_026467.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK011262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK151083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 BC058115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 NM_026467.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144946 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106180 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK011262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK151083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 BC058115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 NM_026467.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK007689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK011262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK151083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 BC058115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 NM_026467.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK011262.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK151083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 BC058115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 NM_026467.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK011262.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK151083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 BC058115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 NM_026467.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26596140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106180 - -
          Whole Mouse Genome G4122A A_51_P246903 AK011262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 AK151083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 BC058115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 NM_026467.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 AK007689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 AK011262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 AK151083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 BC058115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 NM_026467.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246903 AK007689.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126003 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122705 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106180 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07467796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07716355 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07551314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596132 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596133 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596134 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596130 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26596126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26596098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07966424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08137508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07815771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07578272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07578270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08181789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08062730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07716356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07879750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08181780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26596147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10586608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10586607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10586606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10586605 - -
          Affymetrix HT MG-430 PM Array Plate 1423254_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1450925_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1435429_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10586604 - -
          Mouse Genome 430 2.0 Array 1423254_x_at - -
          Mouse Genome 430 2.0 Array 1435429_PM_x_at - -
          Mouse Genome 430 2.0 Array 1423254_PM_x_at - -
          Mouse Genome 430 2.0 Array 1435429_x_at - -
          Mouse Genome 430 2.0 Array 1450925_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450925_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67941_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67941_at - -
          Affymetrix Mouse Clariom S Array PSR0900006798.mm.1 TC0900000876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900006798.mm.1 NM_026467.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900006797.mm.1 TC0900000876.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900006797.mm.1 NM_026467.4 Refseq
          Mouse Expression Set 430 1423254_PM_x_at - -
          Mouse Expression Set 430 1423254_x_at - -
          Mouse Expression Set 430 1450925_PM_a_at - -
          Mouse Expression Set 430 1450925_a_at - -
          Mouse Expression Set 430 1435429_x_at - -
          Mouse Expression Set 430 1435429_PM_x_at - -
          Murine Genome U74v2 Set 160389_r_at - -
          Murine Genome U74v2 Set 160389_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1450925_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423254_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1435429_x_at - -
          Mouse Genome 430A 2.0 Array 1450925_a_at - -
          Mouse Genome 430A 2.0 Array 1435429_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1423254_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423254_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423254_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1435429_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1435429_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450925_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450925_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 718537 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 50577 - -
          Illumina mouse-6 v1.0 expression beadchip 50577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2624280 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2624280 - -
          mouse-6 v1.1 (Illumina) scl067941.3_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 50577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067941.3_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2624280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2624280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067941.3_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0