Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835610_gH
          See other SETD3 GE Assays ›
          Gene Symbol:
          Setd3
          Gene Name:
          SET domain containing 3
          Gene Aliases:
          2610102I01Rik, 2610305M23Rik, 6030490B17Rik, D12Ertd771e
          Chromosome Location:
          Chr.12: 108106431 - 108179381 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835610_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Setd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028262.3 NP_082538.2 3 - 4 334 190
          GenBank mRNA AK011993.1 - 3 - 4 314 190
          AK031371.1 - 3 - 4 297 190
          AK146777.1 - 3 - 4 328 190
          AK166570.1 - 3 - 4 266 190
          AY513271.1 - 3 - 4 267 190
          BC016123.1 - 3 - 4 264 4503842 190
          BC019973.1 - 3 - 4 264 4195130 190

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002345.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Setd3

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K36 methylation
          peptidyl-lysine methylation
          methylation
          positive regulation of transcription by RNA polymerase II
          positive regulation of muscle cell differentiation
          histone H3-K4 methylation
          transcription coactivator activity
          methyltransferase activity
          transferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          histone methyltransferase activity (H3-K36 specific)

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200016318.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016318.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016318.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016326.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016326.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016326.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016316.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016316.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016316.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016320.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016320.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016320.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016330.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016330.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016330.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016321.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016321.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016321.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016317.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016317.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016317.mm.1 NM_028262.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016319.mm.1 TC1200002345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016319.mm.1 NM_028262.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016319.mm.1 NM_028262.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 583555 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0