Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839636_g1
          See other CD68 GE Assays ›
          Gene Symbol:
          Cd68
          Gene Name:
          CD68 antigen
          Gene Aliases:
          Lamp4, Scard1, gp110
          Chromosome Location:
          Chr.11: 69664213 - 69666170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839636_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291058.1 NP_001277987.1 5 - 6 956 86
          NR_110993.1 4 - 5 522 86
          GenBank mRNA AK002264.1 - 5 - 6 914 86
          AK170443.1 - 5 - 6 880 86
          AW123877.1 - 1 - 2 86
          BC021637.1 - 5 - 6 897 3981024 86
          X68273.1 - 5 - 6 939 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003055.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s)

          cellular response to organic substance

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067583 NM_009853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961943 AK170443.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961943 BC021637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961943 X68273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961943 AK002264.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395071 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067583 NM_009853.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27896385 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07386231 - -
          Whole Mouse Genome G4122A A_51_P120470 AK170443.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120470 BC021637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120470 X68273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120470 NM_009853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120470 AK002264.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109871 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07386231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09049549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09214838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09214830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09214820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27896391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09049554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09214831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09214838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09214827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27896385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10387538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387544 - -
          Affymetrix HT MG-430 PM Array Plate 1449164_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10387536 - -
          Mouse Genome 430 2.0 Array 1449164_at - -
          Mouse Genome 430 2.0 Array 1449164_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12514_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12514_at - -
          Affymetrix Mouse Clariom S Array PSR1100027933.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027938.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027939.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027931.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027932.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027934.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027935.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027930.mm.1 TC1100003055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100027937.mm.1 TC1100003055.mm.1 MTA TC
          Mouse Expression Set 430 1449164_PM_at - -
          Mouse Expression Set 430 1449164_at - -
          Murine Genome U74v2 Set 103016_PM_s_at - -
          Murine Genome U74v2 Set 162173_PM_i_at - -
          Murine Genome U74v2 Set 103016_s_at - -
          Murine Genome U74v2 Set 162173_i_at - -
          Mouse Genome 430A 2.0 Array 1449164_at - -
          Mouse Genome 430A 2.0 Array 1449164_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449164_at - -
          Affymetrix HT Mouse Genome 430A Array 1449164_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 587786 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105290301 - -
          MouseWG-6 v2 Expression BeadChip 6350450 - -
          Illumina mouse-6 v1.0 expression beadchip 105290301 - -
          Illumina mouse-6 v1.0 expression beadchip 6350450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689785 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689785 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227424 - -
          mouse-6 v1.1 (Illumina) scl40022.6.1_151-S - -
          mouse-6 v1.1 (Illumina) scl0012514.2_91-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350450 - -
          Illumina Sentrix mouse-6.v1 bead array 105290301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0012514.2_91-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105290301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689785 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689785 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline