Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00839967_g1
          See other CCL19 GE Assays ›
          Gene Symbol:
          Ccl19
          Gene Name:
          chemokine (C-C motif) ligand 19
          Gene Aliases:
          CKb11, ELC, MIP3B, Scya19, exodus-3
          Chromosome Location:
          Chr.4: 42754525 - 42756518 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00839967_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011888.2 NP_036018.1 2 - 3 381 79
          GenBank mRNA AF059208.2 - 2 - 3 381 79
          AK144337.1 - 2 - 3 390 79
          BC051472.1 - 371 1349213 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Mouse Chemokine Panel Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl19

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          dendritic cell chemotaxis
          myeloid dendritic cell chemotaxis
          response to prostaglandin E
          positive regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          release of sequestered calcium ion into cytosol
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          response to nitric oxide
          positive regulation of neutrophil chemotaxis
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          chemokine activity
          CCR10 chemokine receptor binding
          chemokine receptor binding
          CCR chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127995 NM_011888.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127995 AF059208.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127995 AK144337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099742 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127991 BC025130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127991 BC051472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127991 NM_011888.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127991 AF059208.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127991 AK144337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127995 NM_011888.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127995 AF059208.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127995 AK144337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127991 BC025130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127991 BC051472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127991 NM_011888.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127991 AF059208.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127991 AK144337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099742 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458258 NM_011888.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458258 AF059208.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458258 AK144337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458262 AK156269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458262 BC025130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458262 BC051472.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458262 NM_011888.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458262 AF059208.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458262 AK156269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458262 BC025130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458262 BC051472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458262 NM_011888.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458262 AF059208.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458258 NM_011888.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458258 AF059208.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458258 AK144337.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132741 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105634 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109132 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504193 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512374 - -
          Affymetrix HT MG-430 PM Array Plate 1449277_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512322 - -
          Affymetrix Mouse Gene 1.1 ST Array 10504159 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512372 - -
          Affymetrix Mouse Gene 1.1 ST Array 10504188 - -
          Affymetrix Mouse Gene 1.1 ST Array 10504132 - -
          Mouse Genome 430 2.0 Array 1449277_at - -
          Mouse Genome 430 2.0 Array 1449277_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400020221.mm.1 TC0400002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020221.mm.1 NM_011888.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020221.mm.1 NM_011888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020106.mm.1 TC0400002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020107.mm.1 TC0400002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020220.mm.1 TC0400002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020220.mm.1 NM_011888.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020220.mm.1 NM_011888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020111.mm.1 TC0400002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020218.mm.1 TC0400002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020218.mm.1 NM_011888.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020218.mm.1 NM_011888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020222.mm.1 TC0400002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020222.mm.1 NM_011888.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020222.mm.1 NM_011888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020110.mm.1 TC0400002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020113.mm.1 TC0400002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020109.mm.1 TC0400002481.mm.1 MTA TC
          Mouse Expression Set 430 1449277_PM_at - -
          Mouse Expression Set 430 1449277_at - -
          Murine Genome U74v2 Set 96950_at - -
          Murine Genome U74v2 Set 96950_PM_at - -
          Mouse Genome 430A 2.0 Array 1449277_PM_at - -
          Mouse Genome 430A 2.0 Array 1449277_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508185 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080487 - -
          Illumina mouse-6 v1.0 expression beadchip 5080487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080487 - -
          mouse-6 v1.1 (Illumina) scl024047.3_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl024047.3_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2836386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2836386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl024047.3_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0