Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00843415_s1
          See other CCR8 GE Assays ›
          Gene Symbol:
          Ccr8
          Gene Name:
          chemokine (C-C motif) receptor 8
          Gene Aliases:
          C-C, C-C CKR-8, CC-CKR-8, CCR-8, CKR-8, Cmkbr8, mCCR8
          Chromosome Location:
          Chr.9: 120092133 - 120094906 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00843415_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007720.2 NP_031746.1 2 1101 113
          GenBank mRNA BC101966.1 - 1 1045 40044931 113
          BC101967.1 - 1 1045 40044932 113
          BC103566.1 - 1 1045 40044933 113
          BC103567.1 - 1 1045 40044935 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr8

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          immune response
          signal transduction
          G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          chemokine-mediated signaling pathway
          G protein-coupled receptor activity
          chemokine receptor activity
          protein binding
          C-C chemokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136348 NM_007720.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136348 BC101966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136348 BC101967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136348 BC103566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136348 BC103567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103097 NM_007720.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103097 BC101966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103097 BC101967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103097 BC103566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103097 BC103567.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870270 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168523 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167511 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142306 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136348 NM_007720.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136348 BC101966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136348 BC101967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136348 BC103566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136348 BC103567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103097 NM_007720.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103097 BC101966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103097 BC101967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103097 BC103566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103097 BC103567.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225380 NM_007720.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225380 BC101966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225380 BC101967.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225380 BC103566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225380 BC103567.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26876072 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168523 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142306 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167511 - -
          Whole Mouse Genome G4122A A_51_P225380 NM_007720.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225380 BC101966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225380 BC101967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225380 BC103566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225380 BC103567.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107295 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168523 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167511 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08003972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08003976 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07900112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07900110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08003971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08003970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26876060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07748327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07744608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08003970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07748325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07631625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08101023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07631934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07748320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07739736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08101029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08003976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07631932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26876079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07956406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590244 - -
          Affymetrix HT MG-430 PM Array Plate 1422291_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590242 - -
          Mouse Genome 430 2.0 Array 1422291_PM_at - -
          Mouse Genome 430 2.0 Array 1422291_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12776_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12776_at - -
          Mouse Expression Set 430 1422291_at - -
          Mouse Expression Set 430 1422291_PM_at - -
          Murine Genome U74v2 Set 99895_at - -
          Murine Genome U74v2 Set 99895_PM_at - -
          Mouse Genome 430A 2.0 Array 1422291_at - -
          Mouse Genome 430A 2.0 Array 1422291_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422291_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 696905 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1580068 - -
          Illumina mouse-6 v1.0 expression beadchip 1580068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724465 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724465 - -
          mouse-6 v1.1 (Illumina) scl36343.2_555-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36343.2_555-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724465 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724465 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0