Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844089_s1
          See other GPR137C GE Assays ›
          Gene Symbol:
          Gpr137c
          Gene Name:
          G protein-coupled receptor 137C
          Gene Aliases:
          6330416L11Rik, Gm908, TM7SF1L2
          Chromosome Location:
          Chr.14: 45219686 - 45282735 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844089_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr137c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK082691.1 - 1 1749 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569513 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086605 NM_027518.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P589763 AY255546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P589763 NM_027518.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P589763 AY255546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P589763 NM_027518.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427981 BC046559.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427981 XM_914045.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427981 XM_989129.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655720 BC042901.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655720 NM_175021.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166195 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112382 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086605 NM_027518.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P589763 AY255546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P589763 NM_027518.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P589763 AY255546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P589763 NM_027518.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29569537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08096995 - -
          Whole Mouse Genome G4122A A_51_P281912 AK082691.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08096995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29569512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414280 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414282 - -
          Affymetrix HT MG-430 PM Array Plate 1430229_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10414278 - -
          Mouse Genome 430 2.0 Array 1430229_PM_at - -
          Mouse Genome 430 2.0 Array 1430229_at - -
          Mouse Expression Set 430 1430229_PM_at - -
          Mouse Expression Set 430 1430229_at - -
          Murine Genome U74v2 Set 141181_PM_at - -
          Murine Genome U74v2 Set 141181_at - -
          Affymetrix HT Mouse Genome 430B Array 1430229_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430229_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 488245 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 105290086 - -
          Illumina mouse-6 v1.0 expression beadchip 6350373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643128 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643128 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212934 - -
          mouse-6 v1.1 (Illumina) scl070713.5_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 105290086 - -
          Illumina Sentrix mouse-6.v1 bead array 6350373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070713.5_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105290086 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212934 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0