Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00847804_gH
          See other TUBB4B GE Assays ›
          Gene Symbol:
          Tubb4b Zfp646
          Gene Name:
          tubulin, beta 4B class IVB
          zinc finger protein 646
          Gene Aliases:
          4930542G03Rik, Tubb2c, Tubb2c1
          6820429M01
          Chromosome Location:
          Chr.2: 25222158 - 25224702 on Build GRCm38
          Chromosome Location:
          Chr.7: 127877036 - 127885996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00847804_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubb4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146116.2 NP_666228.1 4 1341 144
          GenBank mRNA AK002568.1 - 1 791 144
          AK015901.1 - 4 1291 144
          AK076895.1 - 4 1298 144
          AK146587.1 - 4 1297 144
          AK167022.1 - 4 1296 144
          AK194925.1 - 1 114 144
          AK214079.1 - 1 130 144
          BC005547.1 - 4 1273 3585500 144
          BC022919.1 - 4 1280 4221189 144
          BC083319.1 - 4 1283 5699674 144
          CJ309661.1 - 1 171 144
          Gene Symbol: Zfp646
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC098458.1 - 3064 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tubb4b
          Click here to find siRNAs for Gene Zfp646

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          protein polymerization
          nucleotide binding
          double-stranded RNA binding
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070648 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070649 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070655 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2153292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210260 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143530 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2153292 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P598278 BC022919.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P598278 BC083319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P598278 AK167022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P598278 AK146587.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P598278 NM_146116.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070649 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143530 - -
          Whole Mouse Genome G4122A A_52_P598278 BC022919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P598278 BC083319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P598278 AK167022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P598278 AK146587.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P598278 NM_146116.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111694 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114848 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01377957 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01784164 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01784161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01784151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01784150 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070649 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21070648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01398615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01784154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01784162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01398614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480630 - -
          Affymetrix HT MG-430 PM Array Plate 1423642_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456031_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456470_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1438622_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1439416_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1456078_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519488 - -
          Affymetrix Mouse Gene 1.1 ST Array 10480628 - -
          Mouse Genome 430 2.0 Array 1456031_PM_at - -
          Mouse Genome 430 2.0 Array 1438622_x_at - -
          Mouse Genome 430 2.0 Array 1423642_at - -
          Mouse Genome 430 2.0 Array 1456470_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456470_x_at - -
          Mouse Genome 430 2.0 Array 1456031_at - -
          Mouse Genome 430 2.0 Array 1439416_x_at - -
          Mouse Genome 430 2.0 Array 1423642_PM_at - -
          Mouse Genome 430 2.0 Array 1456078_x_at - -
          Mouse Genome 430 2.0 Array 1439416_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456078_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438622_PM_x_at - -
          Mouse Expression Set 430 1456078_x_at - -
          Mouse Expression Set 430 1456470_PM_x_at - -
          Mouse Expression Set 430 1456031_PM_at - -
          Mouse Expression Set 430 1439416_x_at - -
          Mouse Expression Set 430 1438622_PM_x_at - -
          Mouse Expression Set 430 1456078_PM_x_at - -
          Mouse Expression Set 430 1439416_PM_x_at - -
          Mouse Expression Set 430 1438622_x_at - -
          Mouse Expression Set 430 1423642_PM_at - -
          Mouse Expression Set 430 1456031_at - -
          Mouse Expression Set 430 1456470_x_at - -
          Mouse Expression Set 430 1423642_at - -
          Murine Genome U74v2 Set 94016_PM_at - -
          Murine Genome U74v2 Set 94016_at - -
          Mouse Genome 430A 2.0 Array 1438622_x_at - -
          Mouse Genome 430A 2.0 Array 1423642_PM_at - -
          Mouse Genome 430A 2.0 Array 1456470_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1456031_at - -
          Mouse Genome 430A 2.0 Array 1456031_PM_at - -
          Mouse Genome 430A 2.0 Array 1423642_at - -
          Mouse Genome 430A 2.0 Array 1456470_x_at - -
          Mouse Genome 430A 2.0 Array 1456078_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438622_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439416_x_at - -
          Mouse Genome 430A 2.0 Array 1456078_x_at - -
          Mouse Genome 430A 2.0 Array 1439416_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423642_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423642_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3830672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702061 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702061 - -
          mouse-6 v1.1 (Illumina) scl0227613.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0227613.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702061 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2702061 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0227613.1_83-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0