Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00848037_g1
          See other TAF13 GE Assays ›
          Gene Symbol:
          Taf13
          Gene Name:
          TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          1810004N01Rik, 2010309N11Rik, AI847295, TAFII18
          Chromosome Location:
          Chr.3: 108571699 - 108582068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00848037_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taf13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025444.2 NP_079720.1 4 651 152
          GenBank mRNA AK007339.1 - 4 652 152
          AK076595.1 - 651 152
          AK160171.1 - 4 651 152
          BC021447.1 - 4 663 5050736 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001149.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taf13

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          DNA binding
          transcription coregulator activity
          protein binding
          protein C-terminus binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413427 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108727 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152878 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141280 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098771 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120608 AK007339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120608 AK076595.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120608 AK160171.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120608 BC021447.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P120608 NM_025444.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413433 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141280 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05025546 - -
          Whole Mouse Genome G4122A A_51_P120608 AK007339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120608 AK076595.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120608 AK160171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120608 BC021447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120608 NM_025444.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140625 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02656880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02656883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413427 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02448108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02296394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02656883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02917273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02463190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02769652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02583431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02583432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02359219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02710312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10495341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375974 - -
          Affymetrix HT MG-430 PM Array Plate 1448707_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10375973 - -
          Affymetrix Mouse Gene 1.1 ST Array 10495340 - -
          Mouse Genome 430 2.0 Array 1448707_at - -
          Mouse Genome 430 2.0 Array 1448707_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99730_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99730_at - -
          Affymetrix Mouse Clariom S Array PSR0300009359.mm.1 TC0300001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300009359.mm.1 NM_025444.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009359.mm.1 NM_025444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009358.mm.1 TC0300001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300009358.mm.1 NM_025444.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009358.mm.1 NM_025444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009355.mm.1 TC0300001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300009355.mm.1 NM_025444.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009355.mm.1 NM_025444.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009357.mm.1 TC0300001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300009357.mm.1 NM_025444.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009357.mm.1 NM_025444.2 Refseq
          Mouse Expression Set 430 1448707_at - -
          Mouse Expression Set 430 1448707_PM_at - -
          Murine Genome U74v2 Set 167577_PM_at - -
          Murine Genome U74v2 Set 167577_at - -
          Mouse Genome 430A 2.0 Array 1448707_at - -
          Mouse Genome 430A 2.0 Array 1448707_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448707_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448707_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 562210 - -
          ABI Mouse Genome Survey Microarray 892377 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050692 - -
          Illumina mouse-6 v1.0 expression beadchip 4050692 - -
          Illumina mouse-6 v1.0 expression beadchip 100360687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2728830 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2728830 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2471814 - -
          mouse-6 v1.1 (Illumina) scl0099730.2_131-S - -
          mouse-6 v1.1 (Illumina) scl22718.4_567-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050692 - -
          Illumina Sentrix mouse-6.v1 bead array 100360687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0099730.2_131-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050692 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100360687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2728830 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2728830 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2471814 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0099730.2_131-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc