Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00850051_g1
          See other RPS27 GE Assays ›
          Gene Symbol:
          Rps27 Rps27rt
          Gene Name:
          ribosomal protein S27
          ribosomal protein S27, retrogene
          Gene Aliases:
          3200001M24Rik
          ENSMUSG00000050621, Gm9846, Rps27l2
          Chromosome Location:
          Chr.3: 90212667 - 90213648 on Build GRCm38
          Chromosome Location:
          Chr.9: 114982366 - 114982739 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00850051_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027015.4 NP_081291.1 4 277 152
          NR_033727.1 4 506 152
          GenBank mRNA AA036204.1 - 4 431 152
          AK014294.1 - 4 277 152
          AK084058.1 - 4 277 152
          AK088320.1 - 4 280 152
          AK166551.1 - 4 278 152
          BC027649.1 - 4 263 151
          BC037026.1 - 4 277 152
          BC048352.1 - 4 261 5008906 152
          BC055693.1 - 4 267 6408041 152
          Gene Symbol: Rps27rt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190258.1 NP_001177187.1 1 276 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002366.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps27
          Click here to find siRNAs for Gene Rps27rt

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323988 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155438 NM_001190258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155438 BC027649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155438 BC037026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155438 BC048352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155438 BC055693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163622 AK141404.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113094 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158112 NM_001190258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158112 BC027649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158112 BC037026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158112 BC055693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158112 NM_027015.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163622 AK141404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155438 NM_001190258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155438 BC027649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155438 BC037026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155438 BC048352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155438 BC055693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024322 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511199 AK014294.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22323972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113094 - -
          Whole Mouse Genome G4122A A_51_P511199 AK014294.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114787 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03039719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02825893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03039730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02951608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02825871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02825872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22323988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22323946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02358897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02825888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02358896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02825881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02974379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02883832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02501574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02825894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02974380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03051042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02443317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03051043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02825892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02825893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22323947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499750 - -
          Affymetrix HT MG-430 PM Array Plate 1415716_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1460699_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10499748 - -
          Mouse Genome 430 2.0 Array 1415716_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415716_a_at - -
          Mouse Genome 430 2.0 Array 1460699_PM_at - -
          Mouse Genome 430 2.0 Array 1460699_at - -
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 TC0300002366.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 NM_001190258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 NM_027015.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 NM_027015.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 NR_033727.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018345.mm.1 NR_033727.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 TC0300002366.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 NM_001190258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 NM_027015.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 NM_027015.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 NR_033727.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018344.mm.1 NR_033727.1 Refseq
          Mouse Expression Set 430 1460699_at - -
          Mouse Expression Set 430 1460699_PM_at - -
          Mouse Expression Set 430 1460699_PM_at - -
          Mouse Expression Set 430 1460699_at - -
          Mouse Expression Set 430 1415716_a_at - -
          Mouse Expression Set 430 1415716_PM_a_at - -
          Mouse Expression Set 430 1415716_PM_a_at - -
          Mouse Expression Set 430 1415716_a_at - -
          Murine Genome U74v2 Set 96301_at - -
          Murine Genome U74v2 Set 96301_PM_at - -
          Murine Genome U74v2 Set 96301_at - -
          Murine Genome U74v2 Set 96301_PM_at - -
          Murine Genome U74v2 Set 96300_PM_f_at - -
          Murine Genome U74v2 Set 96300_f_at - -
          Murine Genome U74v2 Set 96300_PM_f_at - -
          Murine Genome U74v2 Set 96300_f_at - -
          Mouse Genome 430A 2.0 Array 1460699_at - -
          Mouse Genome 430A 2.0 Array 1460699_PM_at - -
          Mouse Genome 430A 2.0 Array 1415716_a_at - -
          Mouse Genome 430A 2.0 Array 1415716_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1460699_PM_at - -
          Mouse Genome 430A 2.0 Array 1460699_at - -
          Affymetrix HT Mouse Genome 430A Array 1415716_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415716_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415716_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415716_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774283 - -
          ABI Mouse Genome Survey Microarray 899117 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5270242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669015 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669015 - -
          mouse-6 v1.1 (Illumina) scl057294.3_33-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl057294.3_33-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669015 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl057294.3_33-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0