Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01032120_g1
          See other HNRNPH3 GE Assays ›
          Gene Symbol:
          Hnrnph3 Aff3
          Gene Name:
          heterogeneous nuclear ribonucleoprotein H3
          AF4/FMR2 family, member 3
          Gene Aliases:
          AA693301, AI666703, Hnrph3
          3222402O04Rik, A730046J16, LAF-4, Laf4
          Chromosome Location:
          Chr.10: 63014650 - 63023889 on Build GRCm38
          Chromosome Location:
          Chr.1: 38175991 - 38627837 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01032120_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnph3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079824.1 NP_001073293.1 2 - 3 260 84
          GenBank mRNA BQ443117.1 - 1 - 2 101 84
          Gene Symbol: Aff3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC094538.1 - 101 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002274.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189027 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189032 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 BU946493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 BC049652.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 NM_027425.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 AK042856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 BU946493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 BC049652.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 NM_027425.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P366653 AK042856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059254 NM_001079824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059254 AI666703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059254 AK142212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059254 NM_027425.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755329 AK153123.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755329 AK165846.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755329 NM_023311.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59917 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141459 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 BU946493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 BC049652.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 NM_027425.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 AK042856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 BU946493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 BC049652.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 NM_027425.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P366653 AK042856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059254 NM_001079824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059254 AI666703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059254 AK142212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059254 NM_027425.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 BU946493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 BC049652.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 NM_027425.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 AK042856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 BU946493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 BC049652.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 NM_027425.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P366653 AK042856.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110948 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666214 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08666215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27189027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27189027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27188991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08893328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08300318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08360866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08546123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27188986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369710 - -
          Affymetrix HT MG-430 PM Array Plate 1455491_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369704 - -
          Mouse Genome 430 2.0 Array 1455491_PM_at - -
          Mouse Genome 430 2.0 Array 1455491_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432467_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432467_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000016314.mm.1 TC1000002274.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016314.mm.1 NM_001079824.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016314.mm.1 NM_001079824.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016324.mm.1 TC1000002274.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016324.mm.1 NM_001079824.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016324.mm.1 NM_001079824.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016315.mm.1 TC1000002274.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016315.mm.1 NM_001079824.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016315.mm.1 NM_001079824.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016325.mm.1 TC1000002274.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016325.mm.1 NM_001079824.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016325.mm.1 NM_001079824.1 Refseq
          Mouse Expression Set 430 1455491_PM_at - -
          Mouse Expression Set 430 1455491_at - -
          Murine Genome U74v2 Set 104231_at - -
          Murine Genome U74v2 Set 104231_PM_at - -
          Mouse Genome 430A 2.0 Array 1455491_PM_at - -
          Mouse Genome 430A 2.0 Array 1455491_at - -
          Affymetrix HT Mouse Genome 430A Array 1455491_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455491_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 427482 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2958912 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2958912 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0