Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01129025_g1
          See other CTSD GE Assays ›
          Gene Symbol:
          Ctsd
          Gene Name:
          cathepsin D
          Gene Aliases:
          CD, CatD
          Chromosome Location:
          Chr.7: 142375916 - 142387870 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01129025_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009983.2 NP_034113.1 7 - 8 1026 84
          GenBank mRNA AK089294.1 - 7 - 8 984 84
          AK150363.1 - 7 - 8 987 84
          AK150364.1 - 7 - 8 984 84
          AK150579.1 - 7 - 8 984 84
          AK150622.1 - 7 - 8 984 84
          AK150636.1 - 7 - 8 984 84
          AK150694.1 - 7 - 8 984 84
          AK150768.1 - 7 - 8 984 84
          AK150946.1 - 7 - 8 1036 84
          AK151256.1 - 7 - 8 984 84
          AK151275.1 - 7 - 8 984 84
          AK151276.1 - 7 - 8 984 84
          AK151374.1 - 7 - 8 984 84
          AK151829.1 - 7 - 8 984 84
          AK151962.1 - 7 - 8 987 84
          AK152036.1 - 7 - 8 997 84
          AK152045.1 - 7 - 8 1037 84
          AK152168.1 - 7 - 8 984 84
          AK152187.1 - 7 - 8 984 84
          AK152188.1 - 7 - 8 984 84
          AK152406.1 - 7 - 8 987 84
          AK152458.1 - 7 - 8 984 84
          AK152567.1 - 7 - 8 984 84
          AK152578.1 - 7 - 8 984 84
          AK152593.1 - 7 - 8 1014 84
          AK152638.1 - 7 - 8 996 84
          AK152680.1 - 7 - 8 984 84
          AK152828.1 - 7 - 8 984 84
          AK152881.1 - 7 - 8 984 84
          AK152892.1 - 7 - 8 984 84
          AK153191.1 - 7 - 8 984 84
          AK153291.1 - 7 - 8 984 84
          AK159097.1 - 7 - 8 984 84
          AK159209.1 - 7 - 8 984 84
          AK159440.1 - 7 - 8 1037 84
          AK159490.1 - 7 - 8 984 84
          AK159512.1 - 7 - 8 1026 84
          AK159580.1 - 7 - 8 984 84
          AK159642.1 - 5 - 6 736 84
          AK159743.1 - 7 - 8 988 84
          AK159896.1 - 7 - 8 984 84
          AK159970.1 - 7 - 8 1037 84
          AK165227.1 - 7 - 8 1054 84
          AK166864.1 - 7 - 8 997 84
          AK166938.1 - 7 - 8 1031 84
          AK167143.1 - 7 - 8 1022 84
          AK167366.1 - 7 - 8 1056 84
          AK167698.1 - 7 - 8 983 84
          AK167750.1 - 7 - 8 997 84
          AK167867.1 - 7 - 8 985 84
          AK169139.1 - 7 - 8 1037 84
          AK171195.1 - 7 - 8 985 84
          AK172583.1 - 7 - 8 985 84
          AK205399.1 - 2 - 3 178 84
          AK213803.1 - 2 - 3 178 84
          BC019682.1 - 3 - 4 381 84
          BC048900.1 - 3 - 4 271 84
          BC053937.1 - 3 - 4 271 84
          BC053940.1 - 3 - 4 271 84
          BC054758.1 - 7 - 8 1056 6413315 84
          BC057931.1 - 7 - 8 1017 5253319 84
          X52886.1 - 7 - 8 1026 84
          X53337.1 - 7 - 8 966 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004565.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC048900.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsd

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          proteolysis
          autophagy
          protein catabolic process
          epithelial tube branching involved in lung morphogenesis
          endopeptidase activity
          aspartic-type endopeptidase activity
          peptidase activity
          hydrolase activity
          peptide binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700038470.mm.1 TC0700004565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038470.mm.1 NM_009983.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038470.mm.1 NM_009983.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0