Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01132394_g1
          See other DXO GE Assays ›
          Gene Symbol:
          Dxo
          Gene Name:
          decapping exoribonuclease
          Gene Aliases:
          Dom3z, NG6
          Chromosome Location:
          Chr.17: 34837019 - 34839186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01132394_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dxo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163770.1 NP_001157242.1 2 - 3 561 75
          NM_033613.2 NP_291091.2 2 - 3 586 75
          GenBank mRNA BC004713.1 - 2 - 3 522 3499698 75
          BX522724.1 - 2 - 3 475 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000646.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dxo

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA catabolic process
          metabolic process
          RNA destabilization
          nuclear mRNA surveillance
          nucleic acid phosphodiester bond hydrolysis
          nucleotide binding
          3'-5'-exoribonuclease activity
          magnesium ion binding
          RNA binding
          mRNA binding
          nuclease activity
          exonuclease activity
          5'-3' exoribonuclease activity
          thiamine-diphosphatase activity
          5'-3' exonuclease activity
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          UDP-2,3-diacylglucosamine hydrolase activity
          bis(5'-nucleosyl)-tetraphosphatase activity
          dATP pyrophosphohydrolase activity
          pyrophosphatase activity
          hydrolase activity
          dihydroneopterin monophosphate phosphatase activity
          dihydroneopterin triphosphate pyrophosphohydrolase activity
          RNA pyrophosphohydrolase activity
          dITP diphosphatase activity
          dTTP diphosphatase activity
          XTP diphosphatase activity
          5'-3' DNA helicase activity
          phosphocholine hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700006368.mm.1 TC1700000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006368.mm.1 NM_033613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006368.mm.1 NM_033613.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006372.mm.1 TC1700000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006372.mm.1 NM_001163770.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006372.mm.1 NM_001163770.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006372.mm.1 NM_033613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006372.mm.1 NM_033613.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006373.mm.1 TC1700000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006373.mm.1 NM_001163770.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006373.mm.1 NM_001163770.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006373.mm.1 NM_033613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006373.mm.1 NM_033613.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006375.mm.1 TC1700000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006375.mm.1 NM_001163770.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006375.mm.1 NM_001163770.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006375.mm.1 NM_033613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006375.mm.1 NM_033613.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0