Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01132622_m1
          See other CYP2R1 GE Assays ›
          Gene Symbol:
          Cyp2r1
          Gene Name:
          cytochrome P450, family 2, subfamily r, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 114550123 - 114565618 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01132622_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2r1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177382.4 NP_796356.2 4 - 5 1347 98
          NR_110407.1 4 - 5 773 98
          NR_110408.1 4 - 5 707 98
          GenBank mRNA AK050453.1 - 4 - 5 697 98
          AY323818.1 - 4 - 5 1347 98
          BC108961.1 - 4 - 5 1347 40046632 98
          BC108962.1 - 4 - 5 771 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004141.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2r1

          Gene Ontology Categories:

          Function(s) Process(es)

          organic acid metabolic process
          xenobiotic metabolic process
          calcitriol biosynthetic process from calciol
          xenobiotic catabolic process
          vitamin D metabolic process
          monooxygenase activity
          iron ion binding
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          vitamin D3 25-hydroxylase activity
          metal ion binding
          cholestanetriol 26-monooxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436572 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436570 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436571 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966755 AY323818.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966755 NM_177382.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2201676 AK050453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2201676 AY323818.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2201676 BC108961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2201676 BC108962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2201676 NM_177382.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852985 AK082067.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109208 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154018 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2201676 AK050453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2201676 AY323818.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2201676 BC108961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2201676 BC108962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2201676 NM_177382.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966755 AY323818.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966755 NM_177382.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118046 AK050453.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118046 AY323818.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118046 BC108961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118046 BC108962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118046 NM_177382.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25436591 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01609352 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154018 - -
          Whole Mouse Genome G4122A A_51_P118046 AK050453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118046 AY323818.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118046 BC108961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118046 BC108962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118046 NM_177382.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113879 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127045 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109208 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154018 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05982387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05982373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05982375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05982377 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05982389 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436572 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436571 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436570 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25436574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25436594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06297792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05982371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06472611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06003827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06015278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06003831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25436600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10567091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567088 - -
          Affymetrix HT MG-430 PM Array Plate 1444138_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567086 - -
          Mouse Genome 430 2.0 Array 1444138_PM_at - -
          Mouse Genome 430 2.0 Array 1444138_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244209_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 244209_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700034318.mm.1 TC0700004141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034318.mm.1 NM_177382.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034318.mm.1 NR_110407.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034318.mm.1 NR_110408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034313.mm.1 TC0700004141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034313.mm.1 NM_177382.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034313.mm.1 NR_110407.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034313.mm.1 NR_110408.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034317.mm.1 TC0700004141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034317.mm.1 NM_177382.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034317.mm.1 NR_110407.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034317.mm.1 NR_110408.1 Refseq
          Affymetrix HT Mouse Genome 430B Array 1444138_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448630 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2360010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248189 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248189 - -
          mouse-6 v1.1 (Illumina) scl30785.6.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30785.6.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248189 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248189 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0