Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026521.4 | NP_080797.1 | 3 - 4 | 523 | 80 | |
GenBank mRNA | AB041652.1 | - | 3 - 4 | 466 | 80 | |
AK004076.1 | - | 3 - 4 | 322 | 80 | ||
AK032231.1 | - | 3 - 4 | 409 | 80 | ||
AK082392.1 | - | 3 - 4 | 427 | 80 | ||
AK132053.1 | - | 3 - 4 | 440 | 80 | ||
AK145319.1 | - | 3 - 4 | 401 | 80 | ||
AK166530.1 | - | 3 - 4 | 401 | 80 | ||
AK167791.1 | - | 3 - 4 | 511 | 80 | ||
AK167897.1 | - | 3 - 4 | 440 | 80 | ||
AK168303.1 | - | 3 - 4 | 440 | 80 | ||
AK168958.1 | - | 3 - 4 | 402 | 80 | ||
BC037600.1 | - | 3 - 4 | 448 | 80 | ||
BC042704.1 | - | 3 - 4 | 414 | 80 | ||
BC052459.1 | - | 3 - 4 | 523 | 80 | ||
BC054356.1 | - | 3 - 4 | 445 | 80 | ||
BC085477.1 | - | 3 - 4 | 407 | 80 | ||
BC147306.1 | - | 3 - 4 | 382 | 9056379 | 80 | |
BC147307.1 | - | 3 - 4 | 382 | 9056367 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |