Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01134495_m1
          See other 9430038I01RIK GE Assays ›
          Gene Symbol:
          9430038I01Rik
          Gene Name:
          RIKEN cDNA 9430038I01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 137375569 - 137411900 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01134495_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430038I01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029886.2 NP_084162.1 1 - 2 120 123
          GenBank mRNA AK020460.1 - 1 - 2 92 123
          AK051763.1 - 1 - 2 96 123
          AK078159.1 - 1 - 2 100 123
          AK171836.1 - 1 - 2 121 123
          BY148312.1 - 1 - 2 120 123
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY137825.1 - 1 - 2 121 Chr. 7 - 137387363 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY718759.1 - 1 - 2 95 Chr. 7 - 137376032 - 137410731 [-] on Build GRCm38 123
          GenBankmRNA BY256574.1 - 1 - 2 109 Chr. 7 - 137376216 - 137410745 [-] on Build GRCm38 123
          GenBankmRNA BY750885.1 - 1 - 2 121 Chr. 7 - 137387120 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY084224.1 - 1 - 2 117 Chr. 7 - 137387370 - 137410752 [-] on Build GRCm38 123
          GenBankmRNA AV507625.1 - 1 - 2 112 Chr. 7 - 137376169 - 137410748 [-] on Build GRCm38 123
          GenBankmRNA BY790695.1 - 1 - 2 121 Chr. 7 - 137387361 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BB657093.1 - 1 - 2 95 Chr. 7 - 137387094 - 137410731 [-] on Build GRCm38 123
          GenBankmRNA BY148574.1 - 1 - 2 121 Chr. 7 - 137387401 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY109032.1 - 1 - 2 95 Chr. 7 - 137376328 - 137410731 [-] on Build GRCm38 123
          GenBankmRNA BY721720.1 - 1 - 2 90 Chr. 7 - 137376106 - 137410723 [-] on Build GRCm38 123
          GenBankmRNA BB870772.1 - 1 - 2 95 Chr. 7 - 137376320 - 137410730 [-] on Build GRCm38 123
          GenBankmRNA BY215928.1 - 1 - 2 121 Chr. 7 - 137387476 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY767268.1 - 1 - 2 121 Chr. 7 - 137387333 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY135215.1 - 1 - 2 104 Chr. 7 - 137376306 - 137410739 [-] on Build GRCm38 123
          GenBankmRNA AA624038.1 - 1 - 2 98 Chr. 7 - 137387370 - 137410736 [-] on Build GRCm38 121
          GenBankmRNA BY219149.1 - 1 - 2 121 Chr. 7 - 137387474 - 137410756 [-] on Build GRCm38 123
          GenBankmRNA BY182548.1 - 1 - 2 127 Chr. 7 - 137387348 - 137410762 [-] on Build GRCm38 123

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream early B cell factor 3 Ebf3 3110018A08Rik,
          O/E-2,
          mKIAA4201
          -64.7kb Sense Chr. 7 - 137193671 - 137322810 on NCBI Build GRCm38
          Downstream glutaredoxin 3 Glrx3 PICOT,
          Txnl2
          26.9kb Anti-sense Chr. 7 - 137437643 - 137468594 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK020460.1, AK078159.1, NM_029886.2 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix