Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01135179_m1
          See other HGF GE Assays ›
          Gene Symbol:
          Hgf
          Gene Name:
          hepatocyte growth factor
          Gene Aliases:
          C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF
          Chromosome Location:
          Chr.5: 16553335 - 16622408 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01135179_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hgf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289458.1 NP_001276387.1 15 - 16 1826 76
          NM_001289459.1 NP_001276388.1 15 - 16 1815 76
          NM_001289461.1 NP_001276390.1 14 - 15 1633 76
          NM_010427.5 NP_034557.3 15 - 16 1798 76
          GenBank mRNA BC119228.1 - 15 - 16 1685 8733977 76
          BC144764.1 - 15 - 16 1697 76
          D10212.1 - 14 - 15 1621 76
          D10213.1 - 14 - 15 1606 76
          S71816.1 - 14 - 15 1621 76
          X72307.1 - 14 - 15 1682 76
          X84046.1 - 14 - 15 1661 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000125.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hgf

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          liver development
          proteolysis
          cell population proliferation
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of cell migration
          positive regulation of myelination
          cellular response to hepatocyte growth factor stimulus
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of angiogenesis
          hepatocyte growth factor receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          myoblast proliferation
          cell chemotaxis
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          positive regulation of neuron projection regeneration
          negative regulation of release of cytochrome c from mitochondria
          negative regulation of hydrogen peroxide-mediated programmed cell death
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of DNA biosynthetic process
          serine-type endopeptidase activity
          growth factor activity
          chemoattractant activity
          identical protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500001004.mm.1 TC0500000125.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001004.mm.1 NM_001289458.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001004.mm.1 NM_001289459.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001004.mm.1 NM_001289461.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001004.mm.1 NM_010427.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0