Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01135365_g1
          See other DUSP22 GE Assays ›
          Gene Symbol:
          Dusp22
          Gene Name:
          dual specificity phosphatase 22
          Gene Aliases:
          JKAP, JSP1, LMW-DSP2, MKPX
          Chromosome Location:
          Chr.13: 30660059 - 30711232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01135365_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037955.4 NP_001033044.1 6 - 7 502 68
          NM_134068.3 NP_598829.1 6 - 7 502 68
          GenBank mRNA AF237619.1 - 6 - 7 462 68
          AK148708.1 - 6 - 7 470 68
          AK149363.1 - 6 - 7 461 68
          BC080718.1 - 6 - 7 452 68
          BC108362.1 - 6 - 7 452 4917645 68
          CX236611.1 - 6 - 7 502 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000336.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dusp22

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          negative regulation of T cell mediated immunity
          protein dephosphorylation
          transforming growth factor beta receptor signaling pathway
          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          regulation of cell population proliferation
          positive regulation of JNK cascade
          negative regulation of T cell receptor signaling pathway
          negative regulation of T cell activation
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity
          hydrolase activity
          phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071092 BC057944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095213 BC108362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095213 BC080718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095213 AK149363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095213 AK148708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095213 AF237619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2436438 BC108362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2436438 BC080718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2436438 AK149363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2436438 AK148708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2436438 NM_001037955.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 AK003818.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 AK005296.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 AK011159.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 AK030567.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 AK161187.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 BC027160.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490846 NM_025442.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2436438 BC108362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2436438 BC080718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2436438 AK149363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2436438 AK148708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2436438 NM_001037955.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095213 BC108362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095213 BC080718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095213 AK149363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095213 AK148708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095213 AF237619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231793 BC108362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231793 BC080718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231793 AK149363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231793 AK148708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231793 BC057944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231793 BC108362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231793 BC080718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231793 AK149363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231793 AK148708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231793 BC057944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378211 AF237619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378211 NM_134068.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378211 AK003947.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129552 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404381 - -
          Affymetrix HT MG-430 PM Array Plate 1448985_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404380 - -
          Mouse Genome 430 2.0 Array 1448985_at - -
          Mouse Genome 430 2.0 Array 1448985_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105352_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105352_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300002046.mm.1 TC1300000336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002046.mm.1 NM_001037955.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002046.mm.1 NM_134068.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002049.mm.1 TC1300000336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002049.mm.1 NM_001037955.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002048.mm.1 TC1300000336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002048.mm.1 NM_001037955.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002047.mm.1 TC1300000336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002047.mm.1 NM_001037955.4 Refseq
          Mouse Expression Set 430 1448985_PM_at - -
          Mouse Expression Set 430 1448985_at - -
          Murine Genome U74v2 Set 139525_PM_at - -
          Murine Genome U74v2 Set 169394_i_at - -
          Murine Genome U74v2 Set 169940_i_at - -
          Murine Genome U74v2 Set 139525_at - -
          Murine Genome U74v2 Set 169940_PM_i_at - -
          Murine Genome U74v2 Set 169394_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1448985_at - -
          Mouse Genome 430A 2.0 Array 1448985_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448985_at - -
          Affymetrix HT Mouse Genome 430A Array 1448985_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550546 - -
          Illumina mouse-6 v1.0 expression beadchip 5550546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162564 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215112 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162564 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215112 - -
          mouse-6 v1.1 (Illumina) scl44949.10.388_19-S - -
          mouse-6 v1.1 (Illumina) GI_83816914-I - -
          Illumina Sentrix mouse-6.v1 bead array 5550546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44949.10.388_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2767996 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162564 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2767996 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162564 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_83816914-I - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0