Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01136053_g1
          See other TDP2 GE Assays ›
          Gene Symbol:
          Tdp2
          Gene Name:
          tyrosyl-DNA phosphodiesterase 2
          Gene Aliases:
          D13Ertd656e, Ttrap
          Chromosome Location:
          Chr.13: 24831647 - 24842153 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01136053_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tdp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019551.2 NP_062424.1 6 - 7 910 131
          GenBank mRNA AJ251328.1 - 6 - 7 884 131
          AK009089.1 - 6 - 7 884 131
          AK167707.1 - 5 - 6 706 131
          BC132511.1 - 6 - 7 873 40130788 131
          BC132513.1 - 6 - 7 873 40130790 131
          BC145334.1 - 5 - 6 702 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000280.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tdp2

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair
          cellular response to DNA damage stimulus
          nucleic acid phosphodiester bond hydrolysis
          magnesium ion binding
          single-stranded DNA binding
          cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
          cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
          cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
          photoreceptor cyclic-nucleotide phosphodiesterase activity
          nuclease activity
          endonuclease activity
          phosphoric diester hydrolase activity
          hydrolase activity
          manganese ion binding
          tyrosyl-RNA phosphodiesterase activity
          7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
          metal ion binding
          inositol phosphosphingolipid phospholipase activity
          inositol phosphorylceramide phospholipase activity
          mannosyl-inositol phosphorylceramide phospholipase activity
          mannosyl-diinositol phosphorylceramide phospholipase activity
          5'-tyrosyl-DNA phosphodiesterase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889297 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AK167707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AK217996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 NM_019551.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AJ251328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AK167707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AK217996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 NM_019551.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169047 AJ251328.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143609 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155313 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AK167707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AK217996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 NM_019551.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AJ251328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AK167707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AK217996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 NM_019551.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169047 AJ251328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AK167707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AK217996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 NM_019551.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AJ251328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AK167707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AK217996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 NM_019551.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169047 AJ251328.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28889233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155313 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02648399 - -
          Whole Mouse Genome G4122A A_51_P169047 AK167707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 AK217996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 NM_019551.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 AJ251328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 AK167707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 AK217996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 NM_019551.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169047 AJ251328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56768 BC145334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56768 AK167707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56768 BC132511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56768 BC132513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56768 NM_019551.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127990 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140013 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10926245 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889209 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889210 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889208 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28889212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28889292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11003669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10926245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10687929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10572231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10918642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10791845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10791840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28889224 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404193 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404194 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404192 - -
          Affymetrix HT MG-430 PM Array Plate 1448706_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404187 - -
          Mouse Genome 430 2.0 Array 1448706_PM_at - -
          Mouse Genome 430 2.0 Array 1448706_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56196_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56196_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300001682.mm.1 TC1300000280.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001682.mm.1 NM_019551.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001682.mm.1 NM_019551.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001684.mm.1 TC1300000280.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001684.mm.1 NM_019551.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001684.mm.1 NM_019551.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001683.mm.1 TC1300000280.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001683.mm.1 NM_019551.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001683.mm.1 NM_019551.2 Refseq
          Mouse Expression Set 430 1448706_PM_at - -
          Mouse Expression Set 430 1448706_at - -
          Murine Genome U74v2 Set 93538_at - -
          Murine Genome U74v2 Set 93538_PM_at - -
          Mouse Genome 430A 2.0 Array 1448706_at - -
          Mouse Genome 430A 2.0 Array 1448706_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448706_at - -
          Affymetrix HT Mouse Genome 430A Array 1448706_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 696382 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770041 - -
          Illumina mouse-6 v1.0 expression beadchip 770041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2514218 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2514218 - -
          mouse-6 v1.1 (Illumina) scl44978.8_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 770041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44978.8_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2514218 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2514218 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0