Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01137769_g1
          See other APOBR GE Assays ›
          Gene Symbol:
          Apobr
          Gene Name:
          apolipoprotein B receptor
          Gene Aliases:
          Apob-48r, Apob48r
          Chromosome Location:
          Chr.7: 126584912 - 126589125 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01137769_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apobr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138310.1 NP_612183.1 3 - 4 2763 87
          GenBank mRNA AF141335.1 - 3 - 4 2763 87
          AK156422.1 - 3 - 4 2831 87
          BC030718.1 - 3 - 4 2771 4482457 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001808.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apobr

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          triglyceride metabolic process
          transport
          lipid transport
          receptor-mediated endocytosis
          steroid metabolic process
          cholesterol metabolic process
          lipoprotein particle receptor activity
          very-low-density lipoprotein particle receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 BC030718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 AK156422.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 AF141335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 BC030718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 AK156422.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 AF141335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823655 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113780 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 BC030718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 AK156422.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 AF141335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 NM_138310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 BC030718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 AK156422.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 AF141335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 BC030718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 AK156422.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 AF141335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 BC030718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 AK156422.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 AF141335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501235 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113780 - -
          Whole Mouse Genome G4122A A_51_P203182 BC030718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 AK156422.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 AF141335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 NM_138310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 BC030718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 AK156422.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 AF141335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203182 NM_138310.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113534 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113780 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06116865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06116887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06116860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06116839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557438 - -
          Affymetrix HT MG-430 PM Array Plate 1420382_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557434 - -
          Mouse Genome 430 2.0 Array 1420382_PM_at - -
          Mouse Genome 430 2.0 Array 1420382_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171504_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171504_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700015414.mm.1 TC0700001808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015414.mm.1 NM_138310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015414.mm.1 NM_138310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015412.mm.1 TC0700001808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015412.mm.1 NM_138310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015412.mm.1 NM_138310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015411.mm.1 TC0700001808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015411.mm.1 NM_138310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015411.mm.1 NM_138310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015413.mm.1 TC0700001808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015413.mm.1 NM_138310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015413.mm.1 NM_138310.1 Refseq
          Mouse Expression Set 430 1420382_PM_at - -
          Mouse Expression Set 430 1420382_at - -
          Mouse Genome 430A 2.0 Array 1420382_at - -
          Mouse Genome 430A 2.0 Array 1420382_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420382_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420382_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930150 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2810070 - -
          Illumina mouse-6 v1.0 expression beadchip 2810070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639734 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639734 - -
          mouse-6 v1.1 (Illumina) scl32060.4.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32060.4.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2888834 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639734 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888834 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639734 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0