Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
TC1600000176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_001114085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_001114085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_001285503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_001285504.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_023317.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001259.mm.1 |
NM_023317.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
TC1600000176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_001114085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_001114085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_001285503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_001285504.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_023317.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001255.mm.1 |
NM_023317.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
TC1600000176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_001114085.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_001114085.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_001285503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_001285504.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_023317.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001258.mm.1 |
NM_023317.2 |
Refseq |