Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01150184_m1
          See other PTTG1IP GE Assays ›
          Gene Symbol:
          Pttg1ip
          Gene Name:
          pituitary tumor-transforming 1 interacting protein
          Gene Aliases:
          1810010L20Rik, AI314311, AU018448, C79540
          Chromosome Location:
          Chr.10: 77581767 - 77598732 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01150184_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pttg1ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145925.2 NP_666037.1 4 - 5 500 128
          GenBank mRNA AK035696.1 - 4 - 5 515 128
          AK075773.1 - 4 - 5 473 128
          AK088813.1 - 4 - 5 537 128
          AK089887.1 - 4 - 5 482 128
          AK098093.1 - 4 - 5 514 128
          AK151639.1 - 4 - 5 482 128
          AK152366.1 - 4 - 5 482 128
          AK159987.1 - 4 - 5 482 128
          AK167972.1 - 4 - 5 512 128
          AK168164.1 - 4 - 5 512 128
          AK170802.1 - 4 - 5 482 128
          AK170971.1 - 4 - 5 515 128
          AK170972.1 - 4 - 5 482 128
          AK171140.1 - 4 - 5 481 128
          BC023961.1 - 4 - 5 555 5323397 128
          BC025533.1 - 4 - 5 524 5250502 128
          BC029144.1 - 4 - 5 496 4945974 128
          BC032184.1 - 4 - 5 519 5310191 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000760.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pttg1ip

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          positive regulation of protein ubiquitination
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of cellular protein catabolic process
          p53 binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000005528.mm.1 TC1000000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005528.mm.1 NM_145925.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005528.mm.1 NM_145925.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005526.mm.1 TC1000000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005526.mm.1 NM_145925.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005526.mm.1 NM_145925.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005524.mm.1 TC1000000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005524.mm.1 NM_145925.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005524.mm.1 NM_145925.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0