Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01153075_m1
          See other D630013N20RIK GE Assays ›
          Gene Symbol:
          D630013N20Rik
          Gene Name:
          RIKEN cDNA D630013N20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 70601049 - 70655963 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01153075_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D630013N20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045291.1 5 - 6 522 66
          NR_045292.1 3 - 4 260 66
          GenBank mRNA AK085342.1 - 3 - 4 261 66
          AK090187.1 - 5 - 6 523 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB660954.1 - 3 - 4 258 Chr. 10 - 70602128 - 70655961 [-] on Build GRCm38 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phytanoyl-CoA hydroxylase interacting protein-like Phyhipl 4921522K17Rik,
          AI267048
          -1.8kb Sense Chr. 10 - 70557686 - 70599291 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930533K18 gene 4930533K18Rik 4933433H14Rik 261.0kb Anti-sense Chr. 10 - 70868621 - 70910761 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002350.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368394 AK011003.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368394 BC104344.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368394 BC104345.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368394 NM_027292.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171864 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07099707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02153645 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07099667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07099650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07099760 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108337 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163460 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07099760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08589426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08241168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08241167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08539570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08955841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08800803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08742123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08937523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08937535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08800799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08939541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08241173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08783649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08389277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08937546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08589416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08289508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08663757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08589418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08937525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08565981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08789202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08644998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08709615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08955830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27231044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27230841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1458948_PM_at - -
          Mouse Genome 430 2.0 Array 1458948_at - -
          Mouse Genome 430 2.0 Array 1458948_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000016708.mm.1 TC1000002350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016708.mm.1 NR_045291.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016708.mm.1 NR_045291.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016708.mm.1 NR_045292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016708.mm.1 NR_045292.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016707.mm.1 TC1000002350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016707.mm.1 NR_045291.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016707.mm.1 NR_045291.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016707.mm.1 NR_045292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016707.mm.1 NR_045292.1 Refseq
          Mouse Expression Set 430 1458948_PM_at - -
          Mouse Expression Set 430 1458948_at - -
          Affymetrix HT Mouse Genome 430B Array 1458948_at - -
          Affymetrix HT Mouse Genome 430B Array 1458948_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 890076 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0