Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289602.1 | NP_001276531.1 | 11 - 12 | 2078 | 91 | |
NM_028707.4 | NP_082983.3 | 11 - 12 | 2084 | 91 | ||
GenBank mRNA | AK018116.1 | - | 6 - 7 | 540 | 91 | |
AK034575.1 | - | 11 - 12 | 1853 | 91 | ||
AK045529.1 | - | 11 - 12 | 1815 | 91 | ||
AK149027.1 | - | 11 - 12 | 1814 | 91 | ||
BC056437.1 | - | 7 - 8 | 583 | 91 | ||
BC062930.1 | - | 10 - 11 | 1665 | 6855293 | 91 | |
BC065053.1 | - | 11 - 12 | 1852 | 5709231 | 91 | |
BC065055.1 | - | 11 - 12 | 1835 | 5720949 | 91 | |
BC066026.1 | - | 11 - 12 | 2084 | 5721455 | 91 | |
BC066036.1 | - | 11 - 12 | 1841 | 6837266 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |