Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01158113_g1
          See other TMEM173 GE Assays ›
          Gene Symbol:
          Tmem173
          Gene Name:
          transmembrane protein 173
          Gene Aliases:
          2610307O08Rik, ERIS, MPYS, Mita, STING
          Chromosome Location:
          Chr.18: 35733678 - 35740554 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01158113_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289592.1 NP_001276521.1 1 - 2 246 73
          NM_028261.1 NP_082537.1 1 - 2 246 73
          GenBank mRNA AK012006.1 - 1 - 2 193 73
          AK077788.1 - 1 - 2 147 73
          AK146284.1 - 1 - 2 119 73
          AK153868.1 - 1 - 2 117 73
          AK158458.1 - 1 - 2 155 73
          AK170724.1 - 1 - 2 224 73
          AK171065.1 - 1 - 2 210 73
          AK171203.1 - 1 - 2 210 73
          AK171612.1 - 1 - 2 177 73
          AX775911.1 - 1 - 2 133 73
          BB871885.1 - 1 - 2 249 73
          DQ910493.1 - 1 - 2 44 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001181.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK012006.1, AK077788.1, AK146284.1, AK153868.1, AK158458.1, AK170724.1, AK171065.1, AK171203.1, AK171612.1, AX775911.1, DQ910493.1, NM_001289592.1, NM_028261.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of innate immune response
          positive regulation of defense response to virus by host
          immune system process
          apoptotic process
          positive regulation of protein binding
          positive regulation of type I interferon production
          interferon-beta production
          cellular response to interferon-beta
          innate immune response
          positive regulation of transcription by RNA polymerase II
          defense response to virus
          cellular response to exogenous dsRNA
          nucleotide binding
          protein binding
          transcription factor binding
          protein kinase binding
          ubiquitin protein ligase binding
          cyclic-di-GMP binding
          identical protein binding
          protein homodimerization activity
          2',3'-cyclic GMP-AMP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_028261.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 503137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0