Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01158119_g1
          See other TMEM173 GE Assays ›
          Gene Symbol:
          Tmem173
          Gene Name:
          transmembrane protein 173
          Gene Aliases:
          2610307O08Rik, ERIS, MPYS, Mita, STING
          Chromosome Location:
          Chr.18: 35733678 - 35740554 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01158119_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289591.1 NP_001276520.1 6 - 7 1188 83
          NM_001289592.1 NP_001276521.1 7 - 8 1155 83
          NM_028261.1 NP_082537.1 7 - 8 1278 83
          GenBank mRNA AK012006.1 - 7 - 8 1225 83
          AK089405.1 - 7 - 8 1256 83
          AK146284.1 - 7 - 8 1151 83
          AK153868.1 - 7 - 8 1149 83
          AK157370.1 - 6 - 7 1090 83
          AK158458.1 - 7 - 8 1064 83
          AK170724.1 - 7 - 8 1256 83
          AK171065.1 - 7 - 8 1242 83
          AK171203.1 - 7 - 8 1242 83
          AK171612.1 - 7 - 8 1209 83
          AX775911.1 - 7 - 8 1165 83
          BC027757.1 - 5 - 6 922 83
          BC046640.1 - 6 - 7 1113 5067343 83
          DQ910493.1 - 7 - 8 1076 83
          FJ222242.1 - 5 - 6 940 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001181.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of innate immune response
          positive regulation of defense response to virus by host
          immune system process
          apoptotic process
          positive regulation of protein binding
          positive regulation of type I interferon production
          interferon-beta production
          cellular response to interferon-beta
          innate immune response
          positive regulation of transcription by RNA polymerase II
          defense response to virus
          cellular response to exogenous dsRNA
          nucleotide binding
          protein binding
          transcription factor binding
          protein kinase binding
          ubiquitin protein ligase binding
          cyclic-di-GMP binding
          identical protein binding
          protein homodimerization activity
          2',3'-cyclic GMP-AMP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC027757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC046640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC027757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC046640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13326 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159411 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC027757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC046640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK172314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC027757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC046640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03449407 - -
          Whole Mouse Genome G4122A A_51_P240801 BC027757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC046640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK172314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC027757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC046640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13688687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13640983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14107177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458317 - -
          Affymetrix HT MG-430 PM Array Plate 1427911_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447621_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458314 - -
          Mouse Genome 430 2.0 Array 1427911_at - -
          Mouse Genome 430 2.0 Array 1447621_s_at - -
          Mouse Genome 430 2.0 Array 1427911_PM_at - -
          Mouse Genome 430 2.0 Array 1447621_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72512_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72512_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_028261.1 Refseq
          Mouse Expression Set 430 1427911_PM_at - -
          Mouse Expression Set 430 1427911_at - -
          Mouse Expression Set 430 1447621_PM_s_at - -
          Mouse Expression Set 430 1447621_s_at - -
          Murine Genome U74v2 Set 168500_PM_s_at - -
          Murine Genome U74v2 Set 168500_s_at - -
          Mouse Genome 430A 2.0 Array 1427911_at - -
          Mouse Genome 430A 2.0 Array 1427911_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427911_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427911_at - -
          Affymetrix HT Mouse Genome 430B Array 1447621_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447621_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736197 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736197 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0