Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159249_g1
          See other EZH2 GE Assays ›
          Gene Symbol:
          Ezh2
          Gene Name:
          enhancer of zeste homolog 2 (Drosophila)
          Gene Aliases:
          Enx-1, Enx1h, KMT6, mKIAA4065
          Chromosome Location:
          Chr.6: 47530274 - 47595340 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159249_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ezh2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146689.1 NP_001140161.1 7 - 8 849 103
          NM_007971.2 NP_031997.2 7 - 8 876 103
          GenBank mRNA AK153835.1 - 6 - 7 797 103
          AK157853.1 - 7 - 8 877 103
          AK220174.1 - 7 - 8 831 103
          BC079538.1 - 7 - 8 800 6817366 103
          U52951.1 - 7 - 8 866 103

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ezh2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          regulation of protein phosphorylation
          DNA methylation
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          regulation of gliogenesis
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          histone methylation
          peptidyl-lysine methylation
          cerebellar cortex development
          methylation
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of cell population proliferation
          regulation of circadian rhythm
          positive regulation of MAP kinase activity
          negative regulation of DNA-binding transcription factor activity
          positive regulation of GTPase activity
          negative regulation of epidermal cell differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          rhythmic process
          regulation of neurogenesis
          negative regulation of striated muscle cell differentiation
          cellular response to hydrogen peroxide
          G1 to G0 transition
          histone H3-K27 methylation
          protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of dendrite development
          negative regulation of G1/S transition of mitotic cell cycle
          transcription cis-regulatory region binding
          core promoter sequence-specific DNA binding
          chromatin binding
          RNA binding
          protein binding
          methyltransferase activity
          protein-lysine N-methyltransferase activity
          transferase activity
          histone-lysine N-methyltransferase activity
          chromatin DNA binding
          histone methyltransferase activity
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-85f78c8899-kf4dt:80/100.66.130.62:80.
          git-commit: 7f1fbef35c8161921da6e3861d81379c0ca55d8f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix