Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159973_m1
          See other GDF11 GE Assays ›
          Gene Symbol:
          Gdf11
          Gene Name:
          growth differentiation factor 11
          Gene Aliases:
          BMP-11, Bmp11
          Chromosome Location:
          Chr.10: 128884546 - 128891718 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159973_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdf11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010272.1 NP_034402.1 1 - 2 476 83
          GenBank mRNA AF092734.1 - 1 - 2 221 83

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          metanephros development
          ureteric bud development
          negative regulation of cell population proliferation
          animal organ morphogenesis
          anterior/posterior pattern specification
          positive regulation of pathway-restricted SMAD protein phosphorylation
          spinal cord anterior/posterior patterning
          pancreas development
          growth
          regulation of apoptotic process
          regulation of MAPK cascade
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          cell development
          cell maturation
          camera-type eye morphogenesis
          roof of mouth development
          SMAD protein signal transduction
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543729 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P686760 NM_010272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P686760 NM_010272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25504 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P686760 NM_010272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P686760 AF092734.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P686760 NM_010272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686760 NM_010272.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686760 NM_010272.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02248075 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142224 - -
          Whole Mouse Genome G4122A A_52_P686760 NM_010272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686760 AF092734.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686760 NM_010272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686760 AF092734.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104829 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08311224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08274214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08566983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08311215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08645426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08922665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08340203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08279425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08711300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08645420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08664695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08886473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08231685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08645433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08263866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08450247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08776630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08645427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08498948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08922684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08627529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08312943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08566980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08570875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373586 - -
          Affymetrix HT MG-430 PM Array Plate 1427723_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373583 - -
          Mouse Genome 430 2.0 Array 1427723_at - -
          Mouse Genome 430 2.0 Array 1427723_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14561_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14561_PM_at - -
          Mouse Expression Set 430 1427723_PM_at - -
          Mouse Expression Set 430 1427723_at - -
          Murine Genome U74v2 Set 101814_at - -
          Murine Genome U74v2 Set 101814_PM_at - -
          Mouse Genome 430A 2.0 Array 1427723_at - -
          Mouse Genome 430A 2.0 Array 1427723_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427723_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427723_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 847722 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1780364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223036 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0