Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160166_s1
          See other SMIM10L1 GE Assays ›
          Gene Symbol:
          Smim10l1
          Gene Name:
          small integral membrane protein 10 like 1
          Gene Aliases:
          2700089E24Rik, AI662692
          Chromosome Location:
          Chr.6: 133104914 - 133110899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160166_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim10l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163445.2 NP_001156917.1 1 992 96
          GenBank mRNA AK005205.1 - 1 933 96
          AK031684.1 - 1 972 96
          AK050460.1 - 1 944 96
          AK076076.1 - 1 965 96
          AK170805.1 - 1 944 96
          BC053418.1 - 1 992 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001571.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313337 AK077261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313337 AK077261.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837196 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785062 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313337 AK077261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313337 AK077261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313337 AK077261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313337 AK077261.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05044239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05403522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05671801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05491238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05403532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05403531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P04998164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05295634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24856084 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542265 - -
          Affymetrix HT MG-430 PM Array Plate 1446155_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453208_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460464_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435523_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542264 - -
          Mouse Genome 430 2.0 Array 1460464_at - -
          Mouse Genome 430 2.0 Array 1453208_at - -
          Mouse Genome 430 2.0 Array 1435523_PM_s_at - -
          Mouse Genome 430 2.0 Array 1460464_PM_at - -
          Mouse Genome 430 2.0 Array 1435523_s_at - -
          Mouse Genome 430 2.0 Array 1453208_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600012634.mm.1 TC0600001571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012634.mm.1 NM_001163445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012635.mm.1 TC0600001571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012635.mm.1 NM_001163445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012632.mm.1 TC0600001571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012632.mm.1 NM_001163445.2 Refseq
          Mouse Expression Set 430 1453208_at - -
          Mouse Expression Set 430 1460464_PM_at - -
          Mouse Expression Set 430 1460464_at - -
          Mouse Expression Set 430 1453208_PM_at - -
          Mouse Expression Set 430 1435523_PM_s_at - -
          Mouse Expression Set 430 1446155_PM_at - -
          Mouse Expression Set 430 1446155_at - -
          Mouse Expression Set 430 1435523_s_at - -
          Murine Genome U74v2 Set 100926_at - -
          Murine Genome U74v2 Set 100925_PM_at - -
          Murine Genome U74v2 Set 100926_PM_at - -
          Murine Genome U74v2 Set 100925_at - -
          Murine Genome U74v2 Set 171612_PM_i_at - -
          Murine Genome U74v2 Set 171612_i_at - -
          Mouse Genome 430A 2.0 Array 1453208_at - -
          Mouse Genome 430A 2.0 Array 1460464_at - -
          Mouse Genome 430A 2.0 Array 1460464_PM_at - -
          Mouse Genome 430A 2.0 Array 1453208_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453208_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460464_at - -
          Affymetrix HT Mouse Genome 430A Array 1460464_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453208_at - -
          Affymetrix HT Mouse Genome 430B Array 1435523_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1435523_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 519774 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100430270 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-64bcc5cc5d-xqgf4:80/100.66.130.62:80.
          git-commit: fd21aaec18e3d7b1379e340bd1771dde3ea5c6fc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0