Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01162153_m1
          See other DLX6OS1 GE Assays ›
          Gene Symbol:
          Dlx6os1
          Gene Name:
          distal-less homeobox 6, opposite strand 1
          Gene Aliases:
          A230055N17Rik, Dlx6as1, Evf-1, Evf-2, Evf1, Evf1/2, Evf2, Shhrs, mEvf-1
          Chromosome Location:
          Chr.6: 6820546 - 6869533 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01162153_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlx6os1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK032180.1 - 1 - 2 126 57
          AK038694.1 - 1 - 2 68 57
          AK044034.1 - 1 - 2 85 57

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 AK047127.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 AK077101.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 AK077503.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 AK170524.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 AK171511.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 BC025632.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 BC026926.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 BC031913.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336589 NM_146170.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322805 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656188 AK046793.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656188 AK161890.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656188 BC049167.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656188 BC070410.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656188 NM_175285.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102900 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163938 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165076 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05802528 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05802579 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05802487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05802513 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165076 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163938 - -
          Whole Mouse Genome G4122A A_52_P231012 NR_015388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231012 AK032180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231012 AK032537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231012 AK038694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231012 AK044034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112463 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134126 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163938 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05802487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05871451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05779576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05126171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05779584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05268272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05471667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05471672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05361984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05871461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05398346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05471675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05105583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05550539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05434843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05051730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05100565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05550540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05361983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05779570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05243796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05105584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05779592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05764123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05871463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05871466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05243806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24218631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1438799_PM_at - -
          Mouse Genome 430 2.0 Array 1438799_at - -
          Mouse Genome 430 2.0 Array 1438799_PM_at - -
          Mouse Expression Set 430 1438799_at - -
          Mouse Expression Set 430 1438799_PM_at - -
          Murine Genome U74v2 Set 138093_f_at - -
          Murine Genome U74v2 Set 138093_PM_f_at - -
          Murine Genome U74v2 Set 170818_i_at - -
          Murine Genome U74v2 Set 170818_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1438799_at - -
          Affymetrix HT Mouse Genome 430B Array 1438799_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0