Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01162226_m1
          See other 1700025F24RIK GE Assays ›
          Gene Symbol:
          1700025F24Rik
          Gene Name:
          RIKEN cDNA 1700025F24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 119413444 - 119426864 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01162226_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700025F24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040578.1 3 - 4 506 72
          GenBank mRNA AK006336.1 - 3 - 4 510 72
          AK015773.1 - 3 - 4 485 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715316.1 - 3 - 4 467 Chr. 10 - 119413483 - 119426839 [+] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930477N07 gene 4930477N07Rik -83.3kb Sense Chr. 10 - 119320234 - 119331862 on NCBI Build GRCm38
          Downstream glutamate receptor interacting protein 1 Grip1 4931400F03Rik,
          GRIP,
          eb
          27.0kb Sense Chr. 10 - 119453835 - 120087267 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001465.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144673 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157400 - -
          Whole Mouse Genome G4122A A_52_P598530 AK006336.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P598530 AK015773.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105129 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08371859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08302911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08527889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08527891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08832711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08561265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08215758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27490793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000010396.mm.1 TC1000001465.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000010396.mm.1 NR_040578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010396.mm.1 NR_040578.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0