Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
TC0700004280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001081459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001289538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001289539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001289540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001289541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_001289542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035703.mm.1 |
NM_011363.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
TC0700004280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001081459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001289538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001289539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001289540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001289541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_001289542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035701.mm.1 |
NM_011363.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
TC0700004280.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001081459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001289538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001289539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001289540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001289541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_001289542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035702.mm.1 |
NM_011363.3 |
Refseq |