Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164534_g1
          See other BC024386 GE Assays ›
          Gene Symbol:
          BC024386
          Gene Name:
          cDNA sequence BC023486
          Gene Aliases:
          BC039766
          Chromosome Location:
          Chr.7: 140906064 - 140907979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164534_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC024386
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015583.1 1 - 2 1281 83
          GenBank mRNA BC014806.1 - 1 - 2 1297 83
          BC024386.1 - 3 - 4 392 83
          BC039766.1 - 1 - 2 1301 83
          BC061156.1 - 4 - 5 555 83
          BC087948.1 - 1 - 2 1281 83
          BC100589.1 - 3 - 4 431 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB747982.1 - 1 - 2 174 Chr. 7 - 140907170 - 140907964 [+] on Build GRCm38 83
          GenBankmRNA BI147306.1 - 3 - 4 391 Chr. 7 - 140906043 - 140907979 [+] on Build GRCm38 83
          GenBankmRNA AI391416.1 - 3 - 4 267 Chr. 7 - 140906097 - 140907979 [+] on Build GRCm38 83
          GenBankmRNA AA209058.1 - 3 - 4 266 Chr. 7 - 140906097 - 140907950 [+] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cytochrome c oxidase subunit VIIIb Cox8b Cox8h,
          CoxVIII-H
          -6.7kb Anti-sense Chr. 7 - 140898942 - 140900446 on NCBI Build GRCm38
          Downstream NLR family,
          pyrin domain containing 6
          Nlrp6 AI504961,
          Avr,
          Nalp6,
          Navr,
          Navr/Avr,
          Pypaf5
          13.2kb Sense Chr. 7 - 140920902 - 140929192 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002030.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132979 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700017844.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017844.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017844.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017837.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017837.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017837.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017840.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017840.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017840.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017842.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017842.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017842.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017847.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017847.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017847.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017850.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017850.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017850.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017849.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017849.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017849.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017846.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017846.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017846.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017836.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017836.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017836.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017838.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017838.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017838.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017839.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017839.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017839.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017843.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017843.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017843.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017841.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017841.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017841.mm.1 NR_015583.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017845.mm.1 TC0700002030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700017845.mm.1 NR_015583.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700017845.mm.1 NR_015583.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 432073 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0