Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081058.2 | NP_001074527.1 | 1 - 2 | 1659 | 56 | |
NM_027118.1 | NP_081394.1 | 1 - 2 | 1659 | 56 | ||
GenBank mRNA | AK035493.1 | - | 1 - 2 | 333 | 56 | |
AK083577.1 | - | 1 - 2 | 45 | 56 | ||
AK173265.1 | - | 1 - 2 | 1659 | 56 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |