Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164819_m1
          See other 4930484H19RIK GE Assays ›
          Gene Symbol:
          4930484H19Rik
          Gene Name:
          RIKEN cDNA 4930484H19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 126787739 - 126819722 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164819_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930484H19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015621.1 - 3 - 4 1386 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 378d Mir378d Gm27650,
          mmu-mir-378d
          -78.5kb Sense Chr. 10 - 126710282 - 126710391 on NCBI Build GRCm38
          Downstream XRCC6 binding protein 1 Xrcc6bp1 1110068E08Rik,
          2410012H02Rik
          78.3kb Sense Chr. 10 - 126868427 - 126901711 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2282331 AK015621.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189793 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08880912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08254220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08802763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08819421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08502566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08622020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27532373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431944_PM_at - -
          Mouse Genome 430 2.0 Array 1431944_at - -
          Mouse Genome 430 2.0 Array 1431944_PM_at - -
          Mouse Expression Set 430 1431944_at - -
          Mouse Expression Set 430 1431944_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431944_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431944_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline