Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164820_g1
          See other A330102I10RIK GE Assays ›
          Gene Symbol:
          A330102I10Rik
          Gene Name:
          RIKEN cDNA A330102I10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 29014414 - 29040336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164820_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330102I10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045073.1 1 - 2 136 87
          NR_045074.1 1 - 2 136 87
          NR_045075.1 1 - 2 136 87
          GenBank mRNA AK020728.1 - 1 - 2 101 87
          BY284081.1 - 1 - 2 137 87
          CD769769.1 - 1 - 2 90 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY724424.1 - 1 - 2 100 Chr. 13 - 29016291 - 29028689 [+] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SRY (sex determining region Y)-box 4 Sox4 AA682046,
          Sox-4
          -62.6kb Anti-sense Chr. 13 - 28950716 - 28953682 on NCBI Build GRCm38
          Downstream CDK5 regulatory subunit associated protein 1-like 1 Cdkal1 1190005B03Rik,
          6620401C13Rik
          174.9kb Anti-sense Chr. 13 - 29191751 - 29855684 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000317.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772052 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179590 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10480969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10588348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10601569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11080541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11080539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10480970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10802845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10588356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10823486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10601570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10861842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10995683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10995682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909285 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1458483_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441007_PM_at - -
          Mouse Genome 430 2.0 Array 1441007_PM_at - -
          Mouse Genome 430 2.0 Array 1458483_PM_at - -
          Mouse Genome 430 2.0 Array 1458483_at - -
          Mouse Genome 430 2.0 Array 1441007_at - -
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 TC1300000317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045074.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045074.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001950.mm.1 NR_045075.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 TC1300000317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045074.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045074.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001952.mm.1 NR_045075.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001966.mm.1 TC1300000317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001966.mm.1 NR_045073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001966.mm.1 NR_045073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 TC1300000317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045074.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045074.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001949.mm.1 NR_045075.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001968.mm.1 TC1300000317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001968.mm.1 NR_045073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001968.mm.1 NR_045073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001968.mm.1 NR_045074.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001968.mm.1 NR_045074.1 Refseq
          Mouse Expression Set 430 1458483_at - -
          Mouse Expression Set 430 1441007_PM_at - -
          Mouse Expression Set 430 1458483_PM_at - -
          Mouse Expression Set 430 1441007_at - -
          Murine Genome U74v2 Set 138990_at - -
          Murine Genome U74v2 Set 138990_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458483_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441007_at - -
          Affymetrix HT Mouse Genome 430B Array 1441007_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458483_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-tn8q9:80/100.66.128.91:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0