Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164872_m1
          See other A530046M15RIK GE Assays ›
          Gene Symbol:
          A530046M15Rik
          Gene Name:
          RIKEN cDNA A530046M15 gene
          Gene Aliases:
          A530046M15
          Chromosome Location:
          Chr.13: 15807243 - 15827537 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164872_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530046M15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046131.1 4 - 5 468 63
          GenBank mRNA AK080010.1 - 4 - 5 469 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CO797285.1 - 4 - 5 298 Chr. 13 - 15811287 - 15826891 [+] on Build GRCm38 63
          GenBankmRNA BB636964.1 - 4 - 5 469 Chr. 13 - 15807243 - 15826584 [+] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4933412O06 gene 4933412O06Rik -21.6kb Anti-sense Chr. 13 - 15781664 - 15802630 on NCBI Build GRCm38
          Downstream inhibin beta-A Inhba 180.9kb Sense Chr. 13 - 16007745 - 16027723 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000111.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1700001907.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67846 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07794375 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10535883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10610320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10821832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10641512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10606230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28844302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300000823.mm.1 TC1300000111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000823.mm.1 NR_046131.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000823.mm.1 NR_046131.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000822.mm.1 TC1300000111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000822.mm.1 NR_046131.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000822.mm.1 NR_046131.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 779682 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0