Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600002844.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002844.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002844.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002851.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002851.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002851.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002840.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002840.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002840.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002852.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002852.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002852.mm.1 |
NM_177185.4 |
Refseq |