Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01168492_g1
          See other APOA1 GE Assays ›
          Gene Symbol:
          Apoa1
          Gene Name:
          apolipoprotein A-I
          Gene Aliases:
          Alp-1, Apoa-1, Brp-14, Ltw-1, Lvtw-1, Sep-1, Sep-2, Sep2, apo-AI, apoA-I
          Chromosome Location:
          Chr.9: 46228630 - 46230469 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01168492_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009692.4 NP_033822.2 1 - 2 109 63
          GenBank mRNA AK076187.1 - 1 - 2 32 63
          AK131864.1 - 1 - 2 31 63
          AK149576.1 - 1 - 2 31 63
          AK161536.1 - 1 - 2 31 63
          X64262.1 - 1 - 2 31 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000559.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK076187.1, AK131864.1, AK149576.1, AK161536.1, NM_009692.4, X64262.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoa1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          endothelial cell proliferation
          lipid metabolic process
          phospholipid metabolic process
          phosphatidylcholine biosynthetic process
          cholesterol biosynthetic process
          transport
          lipid transport
          G protein-coupled receptor signaling pathway
          integrin-mediated signaling pathway
          protein localization
          steroid metabolic process
          cholesterol metabolic process
          glucocorticoid metabolic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          negative regulation of very-low-density lipoprotein particle remodeling
          phospholipid transport
          protein oxidation
          peptidyl-methionine modification
          triglyceride catabolic process
          lipid storage
          regulation of intestinal cholesterol absorption
          cholesterol transport
          adrenal gland development
          animal organ regeneration
          axon regeneration
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          negative regulation of heterotypic cell-cell adhesion
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          positive regulation of Rho protein signal transduction
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          blood vessel endothelial cell migration
          reverse cholesterol transport
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          negative regulation of inflammatory response
          protein stabilization
          negative chemotaxis
          positive regulation of lipoprotein lipase activity
          vitamin transport
          positive regulation of hydrolase activity
          negative regulation of hydrolase activity
          positive regulation of transferase activity
          positive regulation of stress fiber assembly
          phospholipid homeostasis
          negative regulation of lipase activity
          negative regulation of cell adhesion molecule production
          negative regulation of response to cytokine stimulus
          triglyceride homeostasis
          cholesterol import
          positive regulation of substrate adhesion-dependent cell spreading
          amyloid-beta binding
          lipid transporter activity
          protein binding
          phospholipid binding
          phospholipid transporter activity
          high-density lipoprotein particle binding
          lipid binding
          cholesterol binding
          enzyme binding
          phosphatidylcholine binding
          apolipoprotein receptor binding
          apolipoprotein A-I receptor binding
          identical protein binding
          chemorepellent activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding
          lipoprotein particle binding
          cholesterol transfer activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900003918.mm.1 TC0900000559.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003918.mm.1 NM_009692.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003912.mm.1 TC0900000559.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003912.mm.1 NM_009692.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 830658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0