Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01168536_s1
          See other H2AFX GE Assays ›
          Gene Symbol:
          H2afx
          Gene Name:
          H2A histone family, member X
          Gene Aliases:
          AW228881, H2A.X, H2ax, Hist5-2ax, gammaH2ax
          Chromosome Location:
          Chr.9: 44334715 - 44336077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01168536_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2afx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010436.2 NP_034566.1 1 1227 69
          GenBank mRNA AK088040.1 - 1 1249 69
          BC005468.1 - 1 1221 3490058 69
          BC010336.1 - 1 1227 3156946 69
          X58069.1 - 1 1229 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2afx

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint signaling
          double-strand break repair via homologous recombination
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          cell cycle
          spermatogenesis
          meiotic cell cycle
          cellular response to gamma radiation
          DNA binding
          damaged DNA binding
          protein binding
          enzyme binding
          histone binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467064 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467067 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467066 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467075 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245275 X58069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245275 X58069.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322159 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245275 X58069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245275 X58069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245275 X58069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245275 X58069.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467077 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06769167 - -
          Whole Mouse Genome G4122A A_51_P245275 X58069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245275 X58069.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07477355 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07477352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07477363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08174016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08174011 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467064 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467066 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08052680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584711 - -
          Affymetrix HT MG-430 PM Array Plate 1416746_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10584710 - -
          Mouse Genome 430 2.0 Array 1416746_at - -
          Mouse Genome 430 2.0 Array 1416746_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15270_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15270_PM_at - -
          Mouse Expression Set 430 1416746_PM_at - -
          Mouse Expression Set 430 1416746_at - -
          Murine Genome U74v2 Set 93019_PM_at - -
          Murine Genome U74v2 Set 93019_at - -
          Mouse Genome 430A 2.0 Array 1416746_PM_at - -
          Mouse Genome 430A 2.0 Array 1416746_at - -
          Affymetrix HT Mouse Genome 430A Array 1416746_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416746_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 907335 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3520082 - -
          Illumina mouse-6 v1.0 expression beadchip 3520082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235873 - -
          mouse-6 v1.1 (Illumina) scl37039.1_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37039.1_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2907878 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2907878 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37039.1_85-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0