Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01169056_gH
          See other DDX21 GE Assays ›
          Gene Symbol:
          Ddx21
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 21
          Gene Aliases:
          AI255159, AL022742, D10Ertd645e, D10Wsu42e
          Chromosome Location:
          Chr.10: 62580247 - 62602298 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01169056_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019553.2 NP_062426.2 1 - 2 173 88
          GenBank mRNA AF159131.3 - 1 - 2 134 88
          AK010281.1 - 1 - 2 53 88
          AK012125.1 - 1 - 2 130 88
          AK145578.1 - 1 - 2 157 88
          AK160095.1 - 1 - 2 147 88
          BC043655.1 - 1 - 2 145 5363948 88
          BC059237.1 - 1 - 2 133 6412782 88
          BC060220.1 - 1 - 2 125 88
          CF182162.1 - 1 - 2 190 88

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK010281.1 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx21

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          transcription, DNA-templated
          rRNA processing
          transcription by RNA polymerase II
          response to virus
          RNA secondary structure unwinding
          response to exogenous dsRNA
          nucleotide binding
          nucleic acid binding
          RNA binding
          RNA helicase activity
          double-stranded RNA binding
          helicase activity
          protein binding
          ATP binding
          hydrolase activity
          rRNA binding
          snoRNA binding
          7SK snRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000016185.mm.1 TC1000002264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016185.mm.1 NM_019553.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016185.mm.1 NM_019553.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016183.mm.1 TC1000002264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016183.mm.1 NM_019553.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016183.mm.1 NM_019553.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016182.mm.1 TC1000002264.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000016182.mm.1 NM_019553.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016182.mm.1 NM_019553.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0