Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01169508_g1
          See other FGF22 GE Assays ›
          Gene Symbol:
          Fgf22
          Gene Name:
          fibroblast growth factor 22
          Gene Aliases:
          2210414E06Rik, FGF-22
          Chromosome Location:
          Chr.10: 79754575 - 79757572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01169508_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023304.1 NP_075793.1 2 - 3 286 73
          GenBank mRNA AB036765.1 - 2 - 3 286 73
          AK008922.1 - 289 73

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf22

          Gene Ontology Categories:

          Function(s) Process(es)

          fibroblast growth factor receptor signaling pathway
          signaling receptor binding
          fibroblast growth factor receptor binding
          protein binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278711 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278712 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006215 NM_023304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006215 AB036765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006215 AK008922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006210 BB823711.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006210 NM_144528.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006210 NR_027505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006210 AK008922.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506851 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142820 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006210 BB823711.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006210 NM_144528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006210 NR_027505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006210 AK008922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006215 NM_023304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006215 AB036765.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006215 AK008922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230773 AK008922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472574 NM_023304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472574 AB036765.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472574 AK008922.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27278724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142820 - -
          Whole Mouse Genome G4122A A_51_P472574 NM_023304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472574 AB036765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P472574 AK008922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230773 AK008922.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135728 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142820 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08398028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278712 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27278711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27278765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08585335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08398014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08398034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27278714 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364492 - -
          Affymetrix HT MG-430 PM Array Plate 1460296_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1450205_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364489 - -
          Mouse Genome 430 2.0 Array 1460296_a_at - -
          Mouse Genome 430 2.0 Array 1460296_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450205_PM_at - -
          Mouse Genome 430 2.0 Array 1450205_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67112_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67112_PM_at - -
          Mouse Expression Set 430 1460296_PM_a_at - -
          Mouse Expression Set 430 1450205_at - -
          Mouse Expression Set 430 1450205_PM_at - -
          Mouse Expression Set 430 1460296_a_at - -
          Mouse Genome 430A 2.0 Array 1450205_PM_at - -
          Mouse Genome 430A 2.0 Array 1450205_at - -
          Mouse Genome 430A 2.0 Array 1460296_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1460296_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450205_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460296_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1460296_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450205_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230735 - -
          Illumina mouse-6 v1.0 expression beadchip 4230735 - -
          Illumina mouse-6 v1.0 expression beadchip 6040450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604814 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245714 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245714 - -
          mouse-6 v1.1 (Illumina) scl067112.1_18-S - -
          mouse-6 v1.1 (Illumina) scl0067112.1_220-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040450 - -
          Illumina Sentrix mouse-6.v1 bead array 4230735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067112.1_18-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067112.1_220-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245714 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc